MilliporeSigma
Catalytic selective deuteration of halo(hetero)arenes.
Organic & biomolecular chemistry (2016-02-24)
Manojkumar Janni, S Peruncheralathan
ABSTRACT
Deuterium labeled aromatic and heteroaromatic compounds are synthesized in good to excellent yields with >98% deuterium purity via palladium catalyzed deuterodehalogenation reaction using commercially available and inexpensive reagents. Selective deuteration of bromoaniline is also demonstrated without H/D exchange in an amino N-H group.
Sigma-Aldrich
Tri(o-tolyl)phosphine, 97%
Sigma-Aldrich
Poly(dimethylsiloxane), viscosity 1.0 cSt (25 °C)