  • Search Results
  • Defects in TRPM7 channel function deregulate thrombopoiesis through altered cellular Mg(2+) homeostasis and cytoskeletal architecture.
Defects in TRPM7 channel function deregulate thrombopoiesis through altered cellular Mg(2+) homeostasis and cytoskeletal architecture.
Nature communications (2016-03-30)
Simon Stritt, Paquita Nurden, Remi Favier, Marie Favier, Silvia Ferioli, Sanjeev K Gotru, Judith M M van Eeuwijk, Harald Schulze, Alan T Nurden, Michele P Lambert, Ernest Turro, Stephanie Burger-Stritt, Masayuki Matsushita, Lorenz Mittermeier, Paola Ballerini, Susanna Zierler, Michael A Laffan, Vladimir Chubanov, Thomas Gudermann, Bernhard Nieswandt, Attila Braun
ABSTRACT
Mg(2+) plays a vital role in platelet function, but despite implications for life-threatening conditions such as stroke or myocardial infarction, the mechanisms controlling [Mg(2+)]i in megakaryocytes (MKs) and platelets are largely unknown. Transient receptor potential melastatin-like 7 channel (TRPM7) is a ubiquitous, constitutively active cation channel with a cytosolic α-kinase domain that is critical for embryonic development and cell survival. Here we report that impaired channel function of TRPM7 in MKs causes macrothrombocytopenia in mice (Trpm7(fl/fl-Pf4Cre)) and likely in several members of a human pedigree that, in addition, suffer from atrial fibrillation. The defect in platelet biogenesis is mainly caused by cytoskeletal alterations resulting in impaired proplatelet formation by Trpm7(fl/fl-Pf4Cre) MKs, which is rescued by Mg(2+) supplementation or chemical inhibition of non-muscle myosin IIA heavy chain activity. Collectively, our findings reveal that TRPM7 dysfunction may cause macrothrombocytopenia in humans and mice.
Product Description
Roche
Thrombin, from human plasma
Sigma-Aldrich
Anti-Tubulin Antibody, Detyrosinated, Chemicon®, from rabbit
Sigma-Aldrich
Anti-beta-Actin antibody, Rabbit monoclonal, recombinant, expressed in proprietary host, clone SP124, affinity isolated antibody
Sigma-Aldrich
Prostaglandin I2 sodium salt, ≥96% (HPLC), synthetic, powder
Sigma-Aldrich
Phalloidin, Fluorescein Isothiocyanate Labeled, sequence Amanita phalloides(synthetic: peptide sequence)
Sigma-Aldrich
Fibrinogen from human plasma, 35-65% protein (≥90% of protein is clottable).
Sigma-Aldrich
Hematoxylin Solution, Mayer′s
Sigma-Aldrich
Fluoroshield, histology mounting medium
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, fatty acid free, essentially globulin free, pH 7, ≥98%
Sigma-Aldrich
1,2-Dilinoleoyl-3-palmitoyl-rac-glycerol, ≥95% (TLC), liquid
Sigma-Aldrich
Apyrase from potatoes, ATPase ≥200 units/mg protein, lyophilized powder
Supelco
GAPDH, standard for protein electrophoresis
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Phalloidin–Atto 647N, BioReagent, suitable for fluorescence, ≥80% (HPLC)
Sigma-Aldrich
2-Phenylindole, technical grade, 95%
Sigma-Aldrich
Anti-Human Kappa Light Chains (Bound and Free)−Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Tubulin Antibody, clone YL1/2, clone YL1/2, Chemicon®, from rat