  • Search Results
  • Resistance training for hot flushes in postmenopausal women: Randomized controlled trial protocol.
Resistance training for hot flushes in postmenopausal women: Randomized controlled trial protocol.
Maturitas (2016-02-10)
Emilia Berin, Mats L Hammar, Hanna Lindblom, Lotta Lindh-Åstrand, Anna-Clara E Spetz Holm
ABSTRACT
Hot flushes and night sweats affect 75% of all women after menopause and is a common reason for decreased quality of life in mid-aged women. Hormone therapy is effective in ameliorating symptoms but cannot be used by all women due to contraindications and side effects. Engagement in regular exercise is associated with fewer hot flushes in observational studies, but aerobic exercise has not proven effective in randomized controlled trials. It remains to be determined whether resistance training is effective in reducing hot flushes and improves quality of life in symptomatic postmenopausal women. The aim of this study is to investigate the effect of standardized resistance training on hot flushes and other health parameters in postmenopausal women. This is an open, parallel-group, randomized controlled intervention study conducted in Linköping, Sweden. Sixty symptomatic and sedentary postmenopausal women with a mean of at least four moderate to severe hot flushes per day or 28 per week will be randomized to an exercise intervention or unchanged physical activity (control group). The intervention consists of 15 weeks of standardized resistance training performed three times a week under supervision of a physiotherapist. The primary outcome is hot flush frequency assessed by self-reported hot flush diaries, and the difference in change from baseline to week 15 will be compared between the intervention group and the control group. The intention is that this trial will contribute to the evidence base regarding effective treatment for hot flushes.
Product Description
Supelco
Serotonin, analytical standard