MilliporeSigma
  • Effect of ovine luteinizing hormone (oLH) charge isoforms on VEGF and cAMP production.
Effect of ovine luteinizing hormone (oLH) charge isoforms on VEGF and cAMP production.
Animal reproduction science (2015-10-29)
Arnulfo Montero-Pardo, Daniel Diaz, Aleida Olivares, Everardo González-Padilla, Clara Murcia, Margarita Gómez-Chavarín, Gabriel Gutiérrez-Ospina, Gerardo Perera-Marín
ABSTRACT
Although an increase in VEGF expression and synthesis in association with LH has been established; it is unknown if all LH isoforms act similarly. This study evaluated the production of cAMP and VEGF among LH isoforms in two in vitro bioassays. The LH was obtained from hypophyses and the group of isoforms was isolated by chromatofocusing. cAMP production was assessed using the in vitro bioassay of HEK-293 cells and VEGF production was evaluated in granulosa cells. Immunological activity was measured with a homologous RIA. Immunoactivity and bioactivity for each isoform were compared against a standard, by estimating the IC50 and the EC50. The basic isoforms were more immunoactive than the standard. The neutral and the moderately acidic had an immunological activity similar to the standard. The acidic isoform was the least immunoreactive. cAMP production at the EC50 dose was similar among the basic isoforms, the moderately acidic and the standard; for the neutral and the acidic, the EC50 dose was higher. It was observed that compared with the control, VEGF production at the lowest LH dose was no different in the standard and each isoform. In the intermediate dose, a positive response was caused in the standard and the neutral and basic isoforms. Although the acidic isoform showed a dose-dependent response, it was not significant relative to the control. In conclusion, the basic isoform generated the greatest cAMP and VEGF production, similar to the reference standard, and the acidic the smallest.
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, suitable for RIA, pH 5.2, ≥96%