  • Search Results
  • Thermodynamic properties of 1-naphthol: Mutual validation of experimental and computational results.
Thermodynamic properties of 1-naphthol: Mutual validation of experimental and computational results.
The Journal of Chemical Thermodynamics (2015)
Chirico RD, et al.