  • Search Results
  • Human JC virus small tumour antigen inhibits nucleotide excision repair and sensitises cells to DNA-damaging agents.
Human JC virus small tumour antigen inhibits nucleotide excision repair and sensitises cells to DNA-damaging agents.
Mutagenesis (2015-03-07)
Jau-Ling Huang, Chang-Shen Lin, Chia-Chu Chang, Yu-Ning Lu, Yu-Ling Hsu, Tzyy-Yue Wong, Yu-Fei Wang
ABSTRACT
The human JC virus (JCV) is potentially carcinogenic to humans as a Group 2B carcinogen, and it is ubiquitous in human populations. To investigate whether the small tumour (ST) antigen of the JCV contributes to genomic instability, we established cell lines stably expressing the JCV ST and examined its role in DNA repair. Results from host cell reactivation (HCR) assay revealed that the established cell lines exhibited lower nucleotide excision repair (NER) activity than the vector control cells did. The presence of γ-H2AX, a marker of DNA damage, indicated that the established cell line contained more DNA damage foci compared with vector control cells. Furthermore, the results of clonogenic analyses indicated that the JCV ST-expressing cells were more sensitive than the vector control cells to ultraviolet (UV) irradiation and cisplatin treatment. Micronuclei formation assay revealed that the JCV ST-positive cells presented more chromosomal breakages than did the JCV ST-negative cells, particularly after exposure to DNA-damaging agents. The xeroderma pigmentosum Group D protein, a DNA helicase involved in NER, was downregulated in the JCV ST-positive cells in response to UV irradiation. The effect of the protein phosphatase 2A (PP2A) inhibitor okadaic acid on NER was similar to that of the ST, which is a PP2A-binding protein. Therefore, the deactivation of the PP2A might underlie ST-mediated NER inhibition. The results of this study indicate that exposing JCV ST-positive cells to DNA-damaging agents causes genomic instability, which contributes to carcinogenesis. Our data provide further evidence on the association between the JCV ST and human cancer.
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90%
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium fluoride solution
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Suplatast tosylate, ≥98% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
DL-Alanine, ≥99%, FCC, FG
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline
Sigma-Aldrich
Okadaic acid from Prorocentrum concavum, 92-100% (HPLC)
Sigma-Aldrich
trans-Platinum(II)diammine dichloride
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
DL-Alanine, ≥99% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O