  • Search Results
  • Cellular microRNA-miR-548g-3p modulates the replication of dengue virus.
Cellular microRNA-miR-548g-3p modulates the replication of dengue virus.
The Journal of infection (2014-12-17)
Weitao Wen, Zhenjian He, Qinlong Jing, Yiwen Hu, Cuiji Lin, Rui Zhou, Xiaoqun Wang, Yangfan Su, Jiehao Yuan, Zhenxin Chen, Jie Yuan, Jueheng Wu, Jun Li, Xun Zhu, Mengfeng Li
ABSTRACT
It has been well recognized that microRNA plays a role in the host-pathogen interaction network. The significance of microRNA in the regulation of dengue virus (DENV) replication, however, remains unknown. The objective of our study was to determine the biological function of miR-548g-3p in modulating the replication of dengue virus. Here we report that employment of a microRNA target search algorithm to analyze the 5' untranslated region (5'UTR) consensus sequences of DENV (DENV serotypes 1-4) led to a discovery that miR-548g-3p directly targets the stem loop A promoter element within the 5'UTR, a region essential for DENV replication. Real-time PCR was used to measure the expression levels of miR-548g-3p under DENV infection. We performed overexpression and inhibition assays to test the role of miR-548g-3p on DENV replication. The protein and mRNA levels of interferon were measured by ELISA and real-time PCR respectively. We found that overexpression of miR-548g-3p suppressed multiplication of DENV 1, 2, 3 and 4, and that miR-548g-3p was also found to interfere with DENV translation, thereby suppressing the expression of viral proteins. Our results suggest that miR-548g-3p directly regulates DENV replication and warrant further study to investigate the feasibility of microRNA-based anti-DENV approaches.
Product Description
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
2-Mercaptoethanol, for molecular biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90%
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Bromophenol Blue
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)