MilliporeSigma
  • Ultradeformable liposomes as multidrug carrier of resveratrol and 5-fluorouracil for their topical delivery.
Ultradeformable liposomes as multidrug carrier of resveratrol and 5-fluorouracil for their topical delivery.
International journal of pharmaceutics (2015-04-23)
Donato Cosco, Donatella Paolino, Jessica Maiuolo, Luisa Di Marzio, Maria Carafa, Cinzia A Ventura, Massimo Fresta
ABSTRACT
Ultradeformable liposomes represent useful formulations able to increase the skin permeation of drug compounds. In this study, resveratrol- and 5-fluorouracil-loaded ultradeformable liposomes were investigated for the potential treatment of non-melanoma skin cancer. The in vitro anticancer activity of ultradeformable liposomes was tested on human skin cancer cells through viability-, cell cycle- and apoptosis-analysis. Furthermore, we tested the percutaneous permeation of ultradeformable liposomes using human stratum corneum and viable epidermis. The co-encapsulation of resveratrol and 5-fluorouracil (multi-drug carrier) in ultradeformable liposomes improved their anticancer activity on skin cancer cells as compared to both the free drug form and the single entrapped agents. These multi-drug ultradeformable liposomes arrest cell proliferation in G1/S, thus modifying the action of 5-fluorouracil and increasing the activity of resveratrol. This effect might depend on the ultradeformable liposomes, which may accumulate in deeper skin layers, thus generating a cutaneous depot from which resveratrol and 5-fluorouracil are gradually released. Resveratrol and 5-fluorouracil co-loaded ultradeformable liposomes could be a new nanomedicine for the treatment of squamous cell carcinoma, i.e., actinic keratosis, Bowen's disease, and keratoacanthoma.
Sigma-Aldrich
Resveratrol, ≥99% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Propidium iodide solution, solution (1.0 mg/ml in water)
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
SAFC
Cholesterol, Plant-Derived, SyntheChol®
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Fluorouracil, meets USP testing specifications
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications