  • Search Results
  • Aminobisphosphonates inhibit dendritic cell-mediated antigen-specific activation of CD1d-restricted iNKT cells.
Aminobisphosphonates inhibit dendritic cell-mediated antigen-specific activation of CD1d-restricted iNKT cells.
Clinical immunology (Orlando, Fla.) (2015-03-23)
Famke L Schneiders, Charlotte M Huijts, Aslihan Mantici, Mica A C Menks, Emmanuel Scotet, Rob Veerhuis, Henk M W Verheul, Tanja D de Gruijl, Hans J van der Vliet
ABSTRACT
CD1d-restricted invariant natural killer T (iNKT) cells constitute an important immunoregulatory T cell subset that can be activated by the synthetic glycolipid α-galactosylceramide (α-GalCer) and initiate antitumor immune responses. As cancer patients are frequently treated with aminobisphosphonates (NBP), it is relevant to determine possible effects of NBP on CD1d-restricted glycolipid Ag-presentation to iNKT cells. We report a striking reduction of α-GalCer-induced iNKT cell activation by monocyte derived dendritic cells (moDC) upon their exposure to NBP during maturation. We found that production of apolipoprotein E (apoE), which is a known facilitator of trans-membrane transport of exogenously derived glycolipids, was significantly diminished in moDC exposed to NBP. As the inhibitory effect of NBP on iNKT cell activation was alleviated by exogenous apoE, our data indicate that reduced apoE production by antigen presenting cells (APC) through NBP limits glycolipid-induced iNKT cell activation. This should be taken into account in the design of iNKT cell-based anti-cancer therapies.
Sigma-Aldrich
6-Phosphonohexanoic acid, 97%
Sigma-Aldrich
D-(+)-Mannose, wood, ≥99%
Sigma-Aldrich
D-(+)-Mannose, synthetic, ≥99%
Sigma-Aldrich
D-(+)-Mannose, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Streptomycin sulfate salt, powder
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
2-Mercaptoethanol, for molecular biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
D-(+)-Mannose, BioUltra, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, for molecular biology, ≥99.0% (GC)