  • Search Results
  • Developmental changes in polyamines and autophagic marker levels in normal and growth-restricted fetal pigs.
Developmental changes in polyamines and autophagic marker levels in normal and growth-restricted fetal pigs.
Journal of animal science (2015-10-07)
Y H Zhu, G Lin, Z L Dai, T J Zhou, T L Yuan, C P Feng, F Chen, G Y Wu, J J Wang
ABSTRACT
Polyamines are essential for embryonic and fetal survival, growth, and development. Additionally, polyamines may induce autophagy in mammalian cells. However, little is known about the availability of polyamines or autophagy in the porcine conceptus with intrauterine growth restriction (IUGR). The present study was performed to evaluate the developmental changes of polyamine concentrations in IUGR and normal porcine fetuses as well as autophagic marker levels in the fetal intestinal mucosa during the second half of gestation when most fetal growth occurs. Allantoic fluid (ALF), amniotic fluid (AMF), umbilical vein, and the small-intestinal mucosa were obtained from both IUGR and normal fetal pigs at d 60, 90, and 110 of gestation. Concentrations of polyamines in fetal fluids as well as protein abundances of microtubule-associated protein light chain 3B (LC3B), an autophagic marker, in the fetal small-intestinal mucosa were determined. Concentrations of polyamines varied greatly in different fetal compartments and changed substantially with advancing gestation. Concentrations of polyamines in IUGR fetal fluids and the small-intestinal mucosa were markedly different from those in their normal counterparts at d 60 and 90 of gestation, whereas most of the differences were not detected by late (d 110) gestation. Specifically, polyamine levels were lower in the umbilical vein plasma but higher in ALF and AMF from IUGR fetuses. Furthermore, enhanced levels of an autophagic marker were observed in the small-intestinal mucosa of IUGR fetuses throughout mid and late gestation in association with abnormal spermidine levels in fetal plasma. These findings support the notion that enhanced autophagy may be an important survival mechanism in IUGR fetuses. Collectively, our findings provide a new framework for future studies to define the roles for polyamines in the prevention and treatment of IUGR in both human medicine and animal production.
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
1,4-Diaminobutane, 99%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tablet
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Spermidine, ≥99% (GC)
Sigma-Aldrich
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder
Sigma-Aldrich
N-Acetyl-L-cysteine, BioXtra, ≥99% (TLC)
Sigma-Aldrich
Spermine, ≥97%
Sigma-Aldrich
Spermidine, BioReagent, for molecular biology, suitable for cell culture, ≥98%
Sigma-Aldrich
N-Acetyl-L-cysteine, BioReagent, suitable for cell culture
Sigma-Aldrich
Spermine, suitable for cell culture, BioReagent
Sigma-Aldrich
Spermidine, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Spermine, ≥99.0% (GC)