  • Search Results
  • Validating Internal Control Genes for the Accurate Normalization of qPCR Expression Analysis of the Novel Model Plant Setaria viridis.
Validating Internal Control Genes for the Accurate Normalization of qPCR Expression Analysis of the Novel Model Plant Setaria viridis.
PloS one (2015-08-08)
Julia Lambret-Frotté, Leandro C S de Almeida, Stéfanie M de Moura, Flavio L F Souza, Francisco S Linhares, Marcio Alves-Ferreira
ABSTRACT
Employing reference genes to normalize the data generated with quantitative PCR (qPCR) can increase the accuracy and reliability of this method. Previous results have shown that no single housekeeping gene can be universally applied to all experiments. Thus, the identification of a suitable reference gene represents a critical step of any qPCR analysis. Setaria viridis has recently been proposed as a model system for the study of Panicoid grasses, a crop family of major agronomic importance. Therefore, this paper aims to identify suitable S. viridis reference genes that can enhance the analysis of gene expression in this novel model plant. The first aim of this study was the identification of a suitable RNA extraction method that could retrieve a high quality and yield of RNA. After this, two distinct algorithms were used to assess the gene expression of fifteen different candidate genes in eighteen different samples, which were divided into two major datasets, the developmental and the leaf gradient. The best-ranked pair of reference genes from the developmental dataset included genes that encoded a phosphoglucomutase and a folylpolyglutamate synthase; genes that encoded a cullin and the same phosphoglucomutase as above were the most stable genes in the leaf gradient dataset. Additionally, the expression pattern of two target genes, a SvAP3/PI MADS-box transcription factor and the carbon-fixation enzyme PEPC, were assessed to illustrate the reliability of the chosen reference genes. This study has shown that novel reference genes may perform better than traditional housekeeping genes, a phenomenon which has been previously reported. These results illustrate the importance of carefully validating reference gene candidates for each experimental set before employing them as universal standards. Additionally, the robustness of the expression of the target genes may increase the utility of S. viridis as a model for Panicoid grasses.
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
Ethidium bromide solution, BioReagent, for molecular biology, 500 μg/mL in H2O
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
Ethidium bromide solution, BioReagent, for molecular biology, 10 mg/mL in H2O
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
Ethidium bromide solution, for fluorescence, ~1% in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Supelco
Pyridaben, PESTANAL®, analytical standard
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Phenol solution, Saturated with 0.1 M citrate buffer, pH 4.3 ± 0.2, BioReagent, for molecular biology
Sigma-Aldrich
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, for molecular biology
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Ethidium bromide, ~95% (HPLC)
Sigma-Aldrich
Ethidium bromide, BioReagent, for molecular biology, powder
Sigma-Aldrich
Magnesium chloride, BioReagent, suitable for insect cell culture, ≥97.0%
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Phenol, for molecular biology
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Phenol, BioUltra, for molecular biology, TE-saturated, ~73% (T)
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC), crystalline (detached)
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Sigma-Aldrich
Phenol, contains hypophosphorous as stabilizer, loose crystals, ACS reagent, ≥99.0%