PGD2 deficiency exacerbates food antigen-induced mast cell hyperplasia.
Nature communications (2015-07-15)
Tatsuro Nakamura, Shingo Maeda, Kazuhide Horiguchi, Toko Maehara, Kosuke Aritake, Byung-il Choi, Yoichiro Iwakura, Yoshihiro Urade, Takahisa Murata
ABSTRACT
Prostaglandin D2 (PGD2) is a major prostanoid secreted mainly by mast cells. Although PGD2 has been identified as a modulator of allergic inflammation, its precise role remains unclear. Here we investigate the role of PGD2 in food allergy. Oral administration of ovalbumin induces allergic responses in sensitized wild-type (WT) mice. Systemic gene deficiency of haematopoietic PGD synthase (H-PGDS(-/-)) exacerbates all of the manifestations accompanying severe mast cell hyperplasia in the intestine. Morphological studies show that c-kit/FcɛRI-positive WT mast cells strongly express H-PGDS. Transplantation of H-PGDS(-/-) mast cells also aggravates ovalbumin-induced mast cell hyperplasia and allergic symptoms in mast cell null mice. H-PGDS deficiency accelerates the production of SDF-1α and the activity of MMP-9 in the antigen-stimulated intestine. SDF-1α receptor blockade or MMP-9 inhibition relieves the exacerbated mast cell hyperplasia and manifestations observed in H-PGDS(-/-). Thus, PGD2 deficiency results in food antigen-induced mast cell hyperplasia.
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Nitrogen, ≥99.998%
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Thymine 1-β-D-arabinofuranoside, ≥99%