  • Search Results
  • The role of nanostructures and hydrophilicity in osseointegration: In-vitro protein-adsorption and blood-interaction studies.
The role of nanostructures and hydrophilicity in osseointegration: In-vitro protein-adsorption and blood-interaction studies.
Journal of biomedical materials research. Part A (2015-01-30)
Brigitte S Kopf, Sylvie Ruch, Simon Berner, Nicholas D Spencer, Katharina Maniura-Weber
ABSTRACT
Protein adsorption and blood coagulation play important roles in the early stages of osseointegration and are strongly influenced by surface properties. We present a systematic investigation of the influence of different surface properties on the adsorption of the blood proteins fibrinogen and fibronectin and the degree of early blood coagulation. Experiments on custom-made and commercially available, microroughened hydrophobic titanium (Ti) surfaces (Ti SLA-Hphob ), hydrophilic (Hphil ) microroughened Ti surfaces with nanostructures (Ti SLActive-Hphil NS), and on bimetallic Ti zirconium alloy (TiZr, Roxolid®) samples were performed, to study the biological response in relation to the surface wettability and the presence of nanostructures (NS). Protein adsorption on the different substrates showed a highly significant effect of surface NS. Hydrophilicity alone did not significantly enhance protein adsorption. Overall, the combination of NS and hydrophilicity led to the highest adsorption levels; independent of whether Ti or TiZr were used. Hydrophilicity induced a strong effect on blood coagulation, whereas the effect of NS alone was weak. The combination of both surface characteristics led to early and most pronounced blood-coagulation. Therefore, nanostructured, hydrophilic Ti and TiZr surfaces may perform better in terms of osseointegration due to continuous protein adsorption and the formation of a layer of blood components on the implant surface.
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, anhydrous, denatured
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Hexamethyldisilazane, reagent grade, ≥99%
Sigma-Aldrich
Sodium chloride solution, 5 M
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Hexamethyldisilazane, ReagentPlus®, 99.9%
Sigma-Aldrich
N,O-Bis(trimethylsilyl)acetamide, synthesis grade, ≥95%
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Carbon, mesoporous, average pore diameter 100 Å±10 Å (typical), >99.95% trace metals basis
Sigma-Aldrich
Ethanesulfonic acid, 95%
Sigma-Aldrich
Carbon, mesoporous, nanopowder, <500 nm particle size (DLS), >99.95% trace metals basis
Sigma-Aldrich
Carbon nanofibers, graphitized (iron-free), composed of conical platelets, D × L 100 nm × 20-200 μm
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Carbon, mesoporous, nanopowder, graphitized, <500 nm particle size (DLS), >99.95% trace metals basis
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Carbon nanofibers, graphitized, platelets (conical), >98% carbon basis, D × L 100 nm × 20-200 μm
Sigma-Aldrich
Carbon, mesoporous
Sigma-Aldrich
Carbon nanofibers, pyrolitically stripped, platelets (conical), >98% carbon basis, D × L 100 nm × 20-200 μm
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.