MilliporeSigma
  • Biocompatibility of ferroelectric lithium niobate and the influence of polarization charge on osteoblast proliferation and function.
Biocompatibility of ferroelectric lithium niobate and the influence of polarization charge on osteoblast proliferation and function.
Journal of biomedical materials research. Part A (2014-12-17)
N Craig Carville, Liam Collins, Michele Manzo, Katia Gallo, Bart I Lukasz, Katey K McKayed, Jeremy C Simpson, Brian J Rodriguez
ABSTRACT
In this work, the influence of substrate surface charge on in vitro osteoblast cell proliferation on ferroelectric lithium niobate (LN) crystal surfaces is investigated. LN has a spontaneous polarization along the z-axis and is thus characterized by positive and negative bound polarization charge at the +z and -z surfaces. Biocompatibility of LN was demonstrated via culturing and fluorescence imaging of MC3T3 osteoblast cells for up to 11 days. The cells showed enhanced proliferation rates and improved osteoblast function through mineral formation on the positively and negatively charged LN surfaces compared to electrostatically neutral x-cut LN and a glass cover slip control. These results highlight the potential of LN as a template for investigating the role of charge on cellular processes.
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, suitable for plant cell culture
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
N-Acetyl-D-lactosamine, ≥98%
Sigma-Aldrich
2-Propanol, BioReagent, for molecular biology, ≥99.5%