  • Search Results
  • A newly developed molecularly imprinted polymer on the surface of TiO2 for selective extraction of triazine herbicides residues in maize, water, and soil.
A newly developed molecularly imprinted polymer on the surface of TiO2 for selective extraction of triazine herbicides residues in maize, water, and soil.
Analytical and bioanalytical chemistry (2015-09-28)
Hao Ran Geng, Shan Shan Miao, She Feng Jin, Hong Yang
ABSTRACT
A new surface molecularly imprinted polymer (MIP) based on nano-TiO2 was developed using propazine (Pro) as a template molecule, ethyleneglycol dimethacrylate (EGDMA) as a crosslinker, methacrylic acid (MAA) as a functional monomer, and 2,2'-azobis (isobutyronitrile) (AIBN) as an initiator. Structures of the newly synthesized surface MIPs were characterized by Fourier transmission infrared spectrometry (FT-IR), scanning electron microscope (SEM), transmission electron microscope (TEM), and X-ray diffraction (XRD). The MIP had a good adsorption capacity and high recognition selectivity to propazine. Meanwhile, it exhibited a cross-selectivity for simazine (Sim) and atrazine (Atr). The MIPs were used as a solid phase extraction (SPE) material. Concomitant extraction, purification, and determination of three pesticides (Pro, Sim, and Atr) residues in water, soil, and maize plant and grain samples were performed by MIP-SPE coupled with high performance liquid chromatography (HPLC). The highly selective separation and enrichment of Pro, Atr, and Sim from the complex environmental media can be achieved. Thus, the newly developed technique provides an analytical platform to quantify the trace amount of Pro, Sim, and Atr residues in multi environmental media and food source.
Sigma-Aldrich
Nitric acid, puriss. p.a., reag. ISO, reag. Ph. Eur., for determinations with dithizone, ≥65%
Sigma-Aldrich
Nitric acid, red, fuming, HNO3 >90 %
Sigma-Aldrich
Nitric acid, 70%, purified by redistillation, ≥99.999% trace metals basis
Sigma-Aldrich
Methyl acetoacetate, ReagentPlus®, 99%
Sigma-Aldrich
Nitric acid, ACS reagent, ≥90.0%
Sigma-Aldrich
Methyl acetoacetate, Arxada quality, ≥99% (GC)
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 5 mm × 8 in.
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Nitric acid, puriss. p.a., 65.0-67.0%
Sigma-Aldrich
Nitric acid, puriss. p.a., ≥65% (T)
Sigma-Aldrich
Nitric acid, ACS reagent, 70%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Acryloyl chloride, ≥97%, contains ~400 ppm phenothiazine as stabilizer
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Acryloyl chloride, 97.0%, contains <210 ppm MEHQ as stabilizer
Sigma-Aldrich
Triethoxysilane, 95%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, NMR reference standard