  • Search Results
  • Au@pNIPAM SERRS Tags for Multiplex Immunophenotyping Cellular Receptors and Imaging Tumor Cells.
Au@pNIPAM SERRS Tags for Multiplex Immunophenotyping Cellular Receptors and Imaging Tumor Cells.
Small (Weinheim an der Bergstrasse, Germany) (2015-05-06)
Gustavo Bodelón, Verónica Montes-García, Cristina Fernández-López, Isabel Pastoriza-Santos, Jorge Pérez-Juste, Luis M Liz-Marzán
ABSTRACT
Detection technologies employing optically encoded particles have gained much interest toward clinical diagnostics and drug discovery, but the portfolio of available systems is still limited. The fabrication and characterization of highly stable surface-enhanced resonance Raman scattering (SERRS)-encoded colloids for the identification and imaging of proteins expressed in cells are reported. These plasmonic nanostructures are made of gold octahedra coated with poly(N-isopropylacrylamide) microgels and can be readily encoded with Raman active dyes while retaining high colloidal stability in biofluids. A layer-by-layer polyelectrolyte coating is used to seal the outer surface of the encoded particles and to provide a reactive surface for covalent conjugation with antibodies. The targeted multiplexing capabilities of the SERRS tags are demonstrated by the simultaneous detection and imaging of three tumor-associated surface biomarkers: epidermal growth factor receptor (EGFR), epithelial cell adhesion molecule (EpCAM), and homing cell adhesion molecule (CD44) by SERRS spectroscopy. The plasmonic microgels are able to discriminate tumor A431 (EGFR+/EpCAM+/CD44+) and nontumor 3T3 2.2 (EGFR-/EpCAM-/CD44+) cells while cocultured in vitro.
Sigma-Aldrich
Anti-Mouse IgG (H+L), F(ab′)2 fragment, CF488A antibody produced in goat, ~2 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
N-Isopropylacrylamide, ≥99%
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
N,N′-Methylenebisacrylamide, suitable for electrophoresis (after filtration or allowing insolubles to settle)
Sigma-Aldrich
N,N′-Methylenebisacrylamide, powder, for molecular biology, suitable for electrophoresis, ≥99.5%
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
2,2′-Azobis(2-methylpropionamidine) dihydrochloride, granular, 97%
Sigma-Aldrich
Argon, ≥99.998%
Sigma-Aldrich
Anti-EGF Receptor antibody, Mouse monoclonal antibody produced in mouse, clone 225, purified from hybridoma cell culture
Sigma-Aldrich
N,N′-Methylenebis(acrylamide), 99%
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Sigma-Aldrich
N-Isopropylacrylamide, 97%
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
N-Hydroxysulfosuccinimide sodium salt, ≥98% (HPLC)
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Argon-40Ar, 99.95 atom %
Sigma-Aldrich
N,N′-Methylenebisacrylamide solution, suitable for electrophoresis, 2% in H2O
Sigma-Aldrich
Phenylacetic acid, suitable for plant cell culture
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
1,2-Dichloroethane, anhydrous, 99.8%
Sigma-Aldrich
Phenylacetic acid, 99%
Sigma-Aldrich
Phenylacetic acid, ≥99%, FCC, FG