Early diagnostic value of Bcl-3 localization in colorectal cancer.
BMC cancer (2015-05-02)
Karunakar Saamarthy, Sofie Björner, Martin Johansson, Göran Landberg, Ramin Massoumi, Karin Jirström, Katarzyna Chmielarska Masoumi
ABSTRACT
B-cell leukemia 3 (Bcl-3) is a member of the inhibitor of κB family, which regulates a wide range of biological processes by functioning as a transcriptional activator or as a repressor of target genes. Elevated expression, sustained nuclear accumulation, and uncontrolled activation of Bcl-3 causes increased cellular proliferation or survival, dependent on the tissue and type of stimuli. We retrospectively reviewed patients who were diagnosed with colorectal cancer at Skåne University Hospital in Malmö between 1st of January 1990 and 31st of December 1991. Bcl-3 localization in colorectal cancer was assessed by immunohistochemistry on tissue microarray and freshly isolated colon from patients. Correlation between Bcl-3 localization and clinicopathological parameters of the cohort were evaluated using the Spearman rank-order correlation coefficient. In addition, Bcl-3 expression and localization in colon adenocarcinoma cells were analysed by western blot, immunohistochemistry and subcellular fractionation separately. We found that Bcl-3 was mainly localized in the cytoplasm in the tumour tissue isolated from colon cancer patients. Normal colon samples from the same patients showed Bcl-3 localization in the nucleus. In three out of six colon cancer cell lines, we detected elevated levels of Bcl-3. In these cell lines Bcl-3 was accumulated in the cytosol. We confirmed these findings by analysing Bcl-3 localization in a colon tissue micro array consisting of 270 cases. In these samples Bcl-3 localization correlated with the proliferation marker Ki-67, but not with the apoptotic marker Caspase 3. These findings indicate that analysis of the subcellular localization of Bcl-3 could be a potential-early diagnostic marker in colon cancer.
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
SAFC
HEPES
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Trimesic acid, 95%
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Trimethylaluminum, 97%
Sigma-Aldrich
Trimethylaluminum solution, 2.0 M in toluene
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.