  • Search Results
  • An alternative clean-up column for the determination of polychlorinated biphenyls in solid matrices.
An alternative clean-up column for the determination of polychlorinated biphenyls in solid matrices.
Environmental science. Processes & impacts (2015-11-13)
Elizabeth N Ndunda, Vincent O Madadi, Boris Mizaikoff
ABSTRACT
The need for continuous monitoring of polychlorinated biphenyls (PCBs) has necessitated the development of analytical techniques that are sensitive and selective with minimal reagent requirement. In light of this, we developed a column for clean-up of soil and sediment extracts, which is less demanding in terms of the amount of solvent and sorbent. The dual-layer column consists of acidified silica gel and molecularly imprinted polymers (MIPs). MIPs were synthesized via aqueous suspension polymerization using PCB 15 as the dummy template, 4-vinylpyridine as the functional monomer and ethylene glycol dimethacrylate as the cross-linker and the obtained particles characterized via SEM, BET, and batch rebinding assays. Pre-concentration of the spiked real-world water sample using MISPE gave recoveries between 85.2 and 104.4% (RSD < 8.69). On the other hand, the specific dual-layer column designed for clean-up of extracts from complex matrices provided recoveries of 91.6-102.5% (RSD < 4%) for spiked soil, which was comparable to clean-up using acidified silica (70.4-90.5%; RSD < 3.72%) and sulfoxide modified silica (89.7-103.0%; RSD < 13.0%). However, the polymers were reusable maintaining recoveries of 79.8-111.8% after 30 cycles of regeneration and re-use, thereby availing a cost-effective clean-up procedure for continuous monitoring of PCBs. Method detection limits were 0.01-0.08 ng g(-1) and 0.002-0.01 ng mL(-1) for solid matrices and water, respectively.
Sigma-Aldrich
Hexane, Laboratory Reagent, ≥95%
Supelco
Dichloromethane solution, certified reference material, 200 μg/mL in methanol
Supelco
Dichloromethane, analytical standard, stabilized
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Nitrogen, ≥99.998%
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Dichloromethane, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
Dichloromethane, biotech. grade, 99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Toluene, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.7% (GC)
Supelco
Toluene, HPLC grade, 99.8%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, Laboratory Reagent, ≥99.3%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%
Supelco
Acetone solution, contains 20.0 % (v/v) acetonitrile, suitable for HPLC
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Supelco
Hexane, suitable for HPLC, ≥95%
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, puriss. p.a., ACS reagent, reag. ISO, ≥99.9% (GC)
Supelco
Acetone solution, certified reference material, 2000 μg/mL in methanol: water (9:1)
Sigma-Aldrich
Hexane, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99% (GC)
Sigma-Aldrich
Dichloromethane, puriss., meets analytical specification of Ph. Eur., NF, ≥99% (GC)
Sigma-Aldrich
Hexane, anhydrous, 95%