MilliporeSigma
Development of Man-rGO for Targeted Eradication of Macrophage Ablation.
Molecular pharmaceutics (2015-07-15)
Byeongtaek Oh, Chi H Lee
ABSTRACT
This study was aimed to develop and evaluate a smart nanosystem that targeted photothermal ablation of inflammatory macrophages in atherosclerotic plaque. Mannosylated-reduced graphene oxide (Man-rGO) was synthesized using three step procedures: (1) preparation of ox-GOs, (2) microwave-assisted synthesis of PEI-rGOs, and (3) mannosylation of PEI-rGO using reductive amination reaction (Man-rGOs). The ζ-potential of Man-rGO that signifies electrophoretic mobility of the charged surface was examined using Zetasizer Nano ZS. The effects of Man-rGO on the cell viability was evaluated using LDH assay and AlamarBlue assay. The targeting efficacy of Man-rGO was assessed using the cellular uptake rate by M2-polarized (i.e., which is induced by IL-4) macrophage. The effects of NOMela loaded in Man-rGO on the enhancement of phagocytosis were evaluated by examining the phagocytic clearance rate of zymosan-FITC particles. The microwave-assisted reduction of GOs was adapted for a facile synthesis of polyethylenimine-reduced GO (PEI-rGO). The mannose functionalization (Man-rGO) of PEI-rGO produced a greater number of amide linkages formed by reductive amination reaction between PEI-rGO and mannose. The ζ-potential of PEI-rGO was +30.6 ± 3.3 mV, whereas that of Man-rGO was down to +13.1 ± 3.8 mV upon interaction with mannose mainly due to the conjugation of mannose on the PEI-rGO surface. Near-infrared (NIR) irradiation increased the temperature of Man-rGO solution to around 45 °C, suggesting that Man-rGO is more potent than ox-GO or rGO in photothermal ablation activity triggered by NIR laser irradiation (808 nm). All testing formulations at the concentrations up to 10 μg/mL exerted less than 10% of membrane disintegration. For AlamarBlue study, more than 90% of cell viability were maintained at the concentrations (up to 10 μg/mL) of all tested formulations. The fluorescent microscopy images of cells after 1 h incubation demonstrated that Man-rGO were mainly accumulated at the subcellular level where the mannose receptors were overexpressed. The cell viability of macrophages significantly decreased upon exposure to Man-rGO irradiated with NIR, but no changes were observed from that of mast cells (for mast cells, 98.3 ± 0.3%; for macrophages, 67.8 ± 1.3%, p < 0.01), indicating that Man-rGO achieved enhanced targetability toward mannose receptor mediated cellular uptake. N-Nitrosomelatonin (NOMela) loaded in macrophage exerted enhanced phagocytic activity. It was concluded that the enhanced photothermal ablation activity of Man-rGO triggered by NIR laser irradiation was mediated through their targetability toward overexpressed mannose receptor, a marker of M2-phenotype of macrophage. The results of this study supported that Man-rGO can serve as an efficient platform for the targeted therapy against atherosclerosis.
Sigma-Aldrich
Ninhydrin, ACS reagent
Sigma-Aldrich
Sodium borohydride solution, 0.5 M in 2-methoxyethyl ether
Sigma-Aldrich
Sodium borohydride solution, ~12 wt. % in 14 M NaOH
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Sigma-Aldrich
D-(+)-Mannose, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
D-(+)-Mannose, BioUltra, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Ninhydrin, suitable for amino acid detection
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
1,3-Dimethyl-2-imidazolidinone, reagent grade
Sigma-Aldrich
Sodium borohydride solution, 2.0 M in triethylene glycol dimethyl ether
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
1,3-Dimethyl-2-imidazolidinone, ≥99.0% (GC)
Sigma-Aldrich
1,3-Dimethyl-2-imidazolidinone, absolute, over molecular sieve (H2O ≤0.04%), ≥99.5% (GC)
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
VenPure® SF, powder
Sigma-Aldrich
D-(+)-Mannose, wood, ≥99%
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N
Sigma-Aldrich
D-(+)-Mannose, synthetic, ≥99%
Sigma-Aldrich
Phosphoric acid-16O4 solution, 70 wt. % in D2O, 99.9 atom % 16O
Sigma-Aldrich
Nitric acid, ACS reagent, ≥90.0%
Sigma-Aldrich
Sodium borohydride, ReagentPlus®, 99%
Sigma-Aldrich
Sodium nitrite, 99.999% trace metals basis
Sigma-Aldrich
Phosphoric acid, crystalline, ≥99.999% trace metals basis
Sigma-Aldrich
Sodium borohydride, purum p.a., ≥96% (gas-volumetric)
Sigma-Aldrich
Phosphoric acid, ≥85 wt. % in H2O, ≥99.999% trace metals basis
Sigma-Aldrich
Sodium borohydride, granular, 99.99% trace metals basis