  • Search Results
  • Regulation of Murine Ovarian Epithelial Carcinoma by Vaccination against the Cytoplasmic Domain of Anti-Müllerian Hormone Receptor II.
Regulation of Murine Ovarian Epithelial Carcinoma by Vaccination against the Cytoplasmic Domain of Anti-Müllerian Hormone Receptor II.
Journal of immunology research (2015-12-01)
Cagri Sakalar, Suparna Mazumder, Justin M Johnson, Cengiz Z Altuntas, Ritika Jaini, Robert Aguilar, Sathyamangla V Naga Prasad, Denise C Connolly, Vincent K Tuohy
ABSTRACT
Anti-Müllerian hormone receptor, type II (AMHR2), is a differentiation protein expressed in 90% of primary epithelial ovarian carcinomas (EOCs), the most deadly gynecologic malignancy. We propose that AMHR2 may serve as a useful target for vaccination against EOC. To this end, we generated the recombinant 399-amino acid cytoplasmic domain of mouse AMHR2 (AMHR2-CD) and tested its efficacy as a vaccine target in inhibiting growth of the ID8 transplantable EOC cell line in C57BL/6 mice and in preventing growth of autochthonous EOCs that occur spontaneously in transgenic mice. We found that AMHR2-CD immunization of C57BL/6 females induced a prominent antigen-specific proinflammatory CD4+ T cell response that resulted in a mild transient autoimmune oophoritis that resolved rapidly with no detectable lingering adverse effects on ovarian function. AMHR2-CD vaccination significantly inhibited ID8 tumor growth when administered either prophylactically or therapeutically, and protection against EOC growth was passively transferred into naive recipients with AMHR2-CD-primed CD4+ T cells but not with primed B cells. In addition, prophylactic AMHR2-CD vaccination of TgMISIIR-TAg transgenic mice significantly inhibited growth of autochthonous EOCs and provided a 41.7% increase in mean overall survival. We conclude that AMHR2-CD vaccination provides effective immunotherapy of EOC with relatively benign autoimmune complications.
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Sodium selenite, 99%
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Sodium selenite, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Nitrilotriacetic acid, Sigma Grade, ≥99%
Sigma-Aldrich
Sodium selenite, γ-irradiated, lyophilized powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Nitrilotriacetic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Nitrilotriacetic acid, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Isopropyl β-D-1-thiogalactopyranoside, ≥99% (TLC)
Sigma-Aldrich
IPTG, ≥99% (TLC), ≤0.1% Dioxane
Sigma-Aldrich
Hydriodic acid, puriss. p.a., ≥67% (T)
Sigma-Aldrich
Hydriodic acid, contains no stabilizer, ACS reagent, 55%
Sigma-Aldrich
Hydriodic acid, contains ≤1.5% hypophosphorous acid as stabilizer, ACS reagent, ≥47.0%
Sigma-Aldrich
Hydriodic acid, 57 wt. % in H2O, distilled, stabilized, 99.95%
Sigma-Aldrich
Hydriodic acid, contains no stabilizer, distilled, 57 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Hydriodic acid, 57 wt. %, distilled, 99.999% trace metals basis
SAFC
Isopropyl β-D-1-thiogalactopyranoside
Sigma-Aldrich
Isopropyl β-D-thiogalactopyranoside solution, ReadyMade IPTG solution for Blue-white screening