MilliporeSigma
  • Protective effect of 14-3-3 antibodies on stressed neuroretinal cells via the mitochondrial apoptosis pathway.
Protective effect of 14-3-3 antibodies on stressed neuroretinal cells via the mitochondrial apoptosis pathway.
BMC ophthalmology (2015-06-28)
Katharina Bell, Corina Wilding, Sebastian Funke, Norbert Pfeiffer, Franz H Grus
ABSTRACT
Previous studies demonstrate changes of autoantibody concentrations against retinal and optic nerve head antigens in the serum of glaucoma patients in comparison to healthy persons. These antibodies belong to the natural autoimmunity. Previous studies showed up regulated, but also significantly down-regulated autoantibody levels. These antibodies have the ability to influence protein profiles of neuroretinal cells and possibly hold neuroprotective potential, as we have been able to demonstrate before. Aim of this study was to analyse the serum and antibody effect of glaucoma patients on neuroretinal cells in more detail and also determine the impact of antibodies found down-regulated in glaucoma patients on the pathogenesis of the neurodegenerative disease glaucoma. Neuroretinal cells (RGC-5) were incubated with serum either from glaucoma patients or healthy controls for 24 h. Mass spectrometric analysis was performed after cell lysis. Furthermore the neuroretinal cells were preincubated with different and concentrations of 14-3-3 antibodies (0.005, 0.1, 0.5, 1, 5 and 10 μg/ml) and then stressed with H2O2, staurosporine or glutamate. Viability tests were performed with crystal violet and ROS tests with DCFH-DA. Antibody location in the cell after antibody incubation was performed with immunocytochemical methods. Additionally mass spectrometric analysis was performed with the cells after antibody incubation. Protein expression analysis with Maldi-Orbitrap MS showed changes in the expression level of regulatory proteins in cells incubated with glaucoma serum, e.g. an up-regulation of 14-3-3 and a down-regulation of Calmodulin. After preincubation of the cells with anti-14-3-3 antibody and stressing the cells, we detected an increase in viability of up to 22 % and a decrease in reactive oxygen species (ROS) of up to 31 %. Proteomic 1 analysis involvement of the mitochondrial apoptosis pathway in this protective effect and immunohistochemical analysis showed an antibody uptake in the cells. We found significant effects of serum antibodies on proteins of neuroretinal cells especially of the mitochondrial apoptosis pathway. Furthermore we detected a protective potential of antibodies down-regulated in glaucoma patients. The changed autoantibodies belong to the natural autoimmunity. We conclude that changes in the natural autoimmunity of patients with glaucoma can negatively impact regulatory functions.
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Water, deuterium-depleted, ≤1 ppm (Deuterium oxide)
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Water, BioPerformance Certified
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
2-Propanol, BioReagent, for molecular biology, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Water-16O, ≥99.94 atom % 16O
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG