MilliporeSigma
Cation-driven Optical Properties of Artificial Luciferases.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry (2015-10-16)
Sung Bae Kim, Simon Miller, Nobuhiro Suzuki, Toshiya Senda, Ryo Nishihara, Koji Suzuki
ABSTRACT
The present study demonstrates cation-driven optical properties of artificial luciferases (ALucs) from copepod luciferases, as an optical readout for bioanalysis. An assignment of the supersecondary structure code (SSC) of ALucs revealed that ALucs carry a helix-loop-helix structure, which appears at the same sites of the EF-hands of typical Ca(2+)-binding proteins. A mutagenesis study shows that the EF-hand-like structure is a pivotal site for enzymatic activity. The effects of 20 kinds of mono- and multivalent cations on ALuc activities were estimated with column-purified ALuc16. High pH values boost the ALuc activities with both the native coelenterazine and an analog called 6-pi-OH-CTZ. Multivalent cations, Ca(II), Mg(II), and Cr(VI), elevate and prolong the ALuc activities, whereas Co(II), Cu(II) and Pb(II) greatly hamper the ALuc activities. Ca(II) greatly prolongs the optical intensities, suggesting a contribution to the structural robustness of ALucs. The inhibitory effect of multivalent cations on the ALuc activities was utilized for creating dose-response curves. The intrinsic cation-driven selectivity and optical intensity of ALucs enable researchers to constitute de novo biosensors for multivalent cations.
Sigma-Aldrich
Imidazole, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%, Redi-Dri, free-flowing
Sigma-Aldrich
Imidazole, for molecular biology, ≥99% (titration), free-flowing, Redi-Dri
Sigma-Aldrich
Boric acid-11B, ≥99 atom % 11B
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%
Sigma-Aldrich
Imidazole, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Imidazole, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Imidazole, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Imidazole, ≥99% (titration), crystalline
Sigma-Aldrich
Potassium phosphate tribasic, reagent grade, ≥98%
Sigma-Aldrich
Imidazole, for molecular biology, ≥99% (titration)
Sigma-Aldrich
Imidazole, ACS reagent, ≥99% (titration)
Sigma-Aldrich
Boric acid, BioXtra, ≥99.5%
Sigma-Aldrich
Boric acid, suitable for electrophoresis, ≥99.5%
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Boric acid, BioReagent, for molecular biology, suitable for cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Citric acid, 99%
Sigma-Aldrich
Boric acid, tablet, 1 g boric acid per tablet
Sigma-Aldrich
Citric acid, anhydrous, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Citric acid, BioUltra, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Imidazole buffer Solution, BioUltra, 1 M in H2O
Sigma-Aldrich
Citric acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
2-Hydroxybutyric acid sodium salt, 97%
Sigma-Aldrich
Boric acid, 99.97% trace metals basis
Sigma-Aldrich
Boric acid, 99.999% trace metals basis
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T)