  • Search Results
  • Lysozyme stability and amyloid fibrillization dependence on Hofmeister anions in acidic pH.
Lysozyme stability and amyloid fibrillization dependence on Hofmeister anions in acidic pH.
Journal of biological inorganic chemistry : JBIC : a publication of the Society of Biological Inorganic Chemistry (2015-06-17)
Slavomíra Poniková, Andrea Antošová, Erna Demjén, Dagmar Sedláková, Jozef Marek, Rastislav Varhač, Zuzana Gažová, Erik Sedlák
ABSTRACT
We have explored an effect of Hofmeister anions, Na2SO4, NaCl, NaBr, NaNO3, NaSCN and NaClO4, on stability and amyloid fibrillization of hen egg white lysozyme at pH 2.7. The stability of the protein was analyzed by differential scanning calorimetry. The Hofmeister effect of the anions was assessed by the parameter dT trs/d[anion] (T trs, transition temperature). We show that dT trs/d[anion] correlates with anion surface tension effects and anion partition coefficients indicating direct interactions between anions and lysozyme. The kinetic of amyloid fibrillization of lysozyme was followed by Thioflavin T (ThT) fluorescence. Negative correlation between dT trs/d[anion] and the nucleation rate of fibrillization in the presence of monovalent anions indicates specific effect of anions on fibrillization rate of lysozyme. The efficiency of monovalent anions to accelerate fibrillization correlates with inverse Hofmeister series. The far-UV circular dichroism spectroscopy and atomic force microscopy findings show that conformational properties of fibrils depend on fibrillization rate. In the presence of sodium chloride, lysozyme forms typical fibrils with elongated structure and with the secondary structure of the β-sheet. On the other hand, in the presence of both chaotropic perchlorate and kosmotropic sulfate anions, the fibrils form clusters with secondary structure of β-turn. Moreover, the acceleration of fibril formation is accompanied by decreased amount of the formed fibrils as indicated by ThT fluorescence. Taken together, our study shows Hofmeister effect of monovalent anions on: (1) lysozyme stability; (2) ability to accelerate nucleation phase of lysozyme fibrillization; (3) amount, and (4) conformational properties of the formed fibrils.
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture