MilliporeSigma
  • Chrysin inhibits diabetic renal tubulointerstitial fibrosis through blocking epithelial to mesenchymal transition.
Chrysin inhibits diabetic renal tubulointerstitial fibrosis through blocking epithelial to mesenchymal transition.
Journal of molecular medicine (Berlin, Germany) (2015-06-13)
Min-Kyung Kang, Sin-Hye Park, Yean-Jung Choi, Daekeun Shin, Young-Hee Kang
ABSTRACT
Renal fibrosis is a crucial event in the pathogenesis of diabetic nephropathy (DN). The process known as epithelial to mesenchymal transition (EMT) contributes to the accumulation of matrix proteins in kidneys, in which renal tubular epithelial cells play an important role in progressive renal fibrosis. The current study investigated that chrysin (5,7-dihydroxyflavone) present in bee propolis and herbs, inhibited renal tubular EMT and tubulointerstitial fibrosis due to chronic hyperglycemia. Human renal proximal tubular epithelial cells (RPTEC) were incubated in media containing 5.5 mM glucose, 27.5 mM mannitol (as an osmotic control), or 33 mM glucose (HG) in the absence and presence of 1-20 μM chrysin for 72 h. Chrysin significantly inhibited high glucose-induced renal EMT through blocking expression of the mesenchymal markers vimentin, α-smooth muscle actin, and fibroblast-specific protein-1 in RPTEC and db/db mice. Chrysin reversed the HG-induced down-regulation of the epithelial marker E-cadherin and the HG-enhanced N-cadherin induction in RPTEC. In addition, chrysin inhibited the production of collagen IV in tubular cells and the deposition of collagen fibers in mouse kidneys. Furthermore, chrysin blocked tubular cell migration concurrent with decreasing matrix metalloproteinase-2 activity, indicating epithelial cell derangement and tubular basement membrane disruption. Chrysin restored the induction of the tight junction proteins Zona occludens protein-1 (ZO-1) and occludin downregulated in diabetic mice. Chrysin inhibited renal tubular EMT-mediated tubulointerstitial fibrosis caused by chronic hyperglycemia. Therefore, chrysin may be a potent renoprotective agent for the treatment of renal fibrosis-associated DN. • Glucose increases renal tubular epithelial induction of vimentin, α-SMA and FSP-1. • Glucose enhances renal EMT by blocking tubular epithelial E-cadherin expression. • Chrysin inhibits tubular EMT-mediated tubulointerstitial fibrosis in mouse kidneys. • Chrysin restores renal tubular induction of ZO-1 and occludin downregulated in diabetic mice. • Chrysin blocks glucose-induced renal tubular cell migration with reducing MMP-2 activity.
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
D-(+)-Glucose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Chrysin, 97%
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
D-Mannitol, ≥98%