  • Search Results
  • All-trans retinoic acid downregulates ALDH1-mediated stemness and inhibits tumour formation in ovarian cancer cells.
All-trans retinoic acid downregulates ALDH1-mediated stemness and inhibits tumour formation in ovarian cancer cells.
Carcinogenesis (2015-03-07)
Ming-Jer Young, Yi-Hui Wu, Wen-Tai Chiu, Tzu-Yu Weng, Yu-Fang Huang, Cheng-Yang Chou
ABSTRACT
Aldehyde dehydrogenase 1 (ALDH1) is a cancer stem-like cell (CSC) marker in human cancers; however, the specific ALDH1-regulated function and its underlying signalling pathways have not been fully demonstrated. Here, we investigated the ALDH1-regulated function and its underlying signalling and tested whether all-trans retinoic acid (ATRA) can suppress ALDH1-regulated tumour behaviour in ovarian cancer cells. By modulating ALDH1 expression using flow cytometry enrichment and exogenous overexpression or knockdown, we showed that the ALDH1 activity is positively correlated with stemness in ovarian cancer cells according to measures such as sphere formation and CSC marker expression as well as tumourigenesis in a mouse xenograft model. The findings indicate that the ALDH1 directly regulates the functions of ovarian cancer cells. We also showed that ALDH1 can regulate the expression of FoxM1 and Notch 1, which are involved in the downstream signalling of ALDH1-mediated biofunctions. Inhibition of FoxM1 by Thiostrepton and of Notch1 by DAPT downregulated the sphere formation ability of cells. ATRA reduced ALDH1 expression, suppressed tumour formation and inhibited sphere formation, cell migration and invasion in ALDH1-abundant ovarian cancer cells. We conclude that ATRA downregulates ALDH1/FoxM1/Notch1 signalling and suppresses tumour formation in ovarian cancer cells.
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
DAPT, ≥98% (HPLC), solid
Sigma-Aldrich
EGF from mouse, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)
Sigma-Aldrich
EGF human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
EGF from mouse, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
hEGF, EGF, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Supelco
Hematoxylin
Sigma-Aldrich
Hematoxylin, certified by the Biological Stain Commission
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
EGF from rat, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Epidermal Growth Factor, human, animal component free, EGF, recombinant, expressed in Escherichia coli, >97% (SDS-PAGE)
Sigma-Aldrich
Propidium iodide solution, solution (1.0 mg/ml in water)