  • Search Results
  • C,N-bipyrazole receptor grafted onto a porous silica surface as a novel adsorbent based polymer hybrid.
C,N-bipyrazole receptor grafted onto a porous silica surface as a novel adsorbent based polymer hybrid.
Talanta (2015-06-17)
Smaail Radi, Ahmed Attayibat, Mohamed El-Massaoudi, Maryse Bacquet, Shehdeh Jodeh, Ismail Warad, Salim S Al-Showiman, Yahia N Mabkhot
ABSTRACT
A simple heterogeneous synthesis of pure adsorbent based polymer hybrid made by condensing a functionalized C,N-bipyrazole with a 3-glycidoxypropyl-trimethoxysilane silylant agent, previously anchored on a silica surface was developed. The formed material (SG2P) was characterized through elemental analysis, FT-IR spectroscopy, (13)C NMR of solid state, scanning electron microscope (SEM), and was studied and evaluated by determination of the surface area using the BET equation, the adsorption and desorption capability using the isotherm of nitrogen and B.J.H. pore sizes. The new material exhibits good thermal stability determined by thermogravimetry curves and good chemical stability was examined in various acidic and buffer solutions (pH 1-7). The binding and adsorption abilities of SG2P were investigated for Hg(2+), Cd(2+), Pb(2+), Zn(2+), K(+), Na(+) and Li(+) cations and compared to the results of classical liquid-liquid extraction with the unbound C,N-bipyrazole compound. The grafting at the surface of silica does not affect complexing properties of the ligand and the SG2P exhibits a high selectivity toward Hg(2+) ion with no complexation being observed towards zinc and alkali metals. The extracted and the complexing cation percentages were determined by atomic absorption measurements.
Sigma-Aldrich
(3-Glycidyloxypropyl)trimethoxysilane, ≥98%