  • Search Results
  • Effective photothermal chemotherapy with docetaxel-loaded gold nanospheres in advanced prostate cancer.
Effective photothermal chemotherapy with docetaxel-loaded gold nanospheres in advanced prostate cancer.
Journal of drug targeting (2015-03-05)
Yanfang Shen, Zhiya Ma, Fei Chen, Qingjian Dong, Qiran Hu, Lingyu Bai, Jing Chen
ABSTRACT
Multifunctional gold nanospheres (MGNs)-loaded with docetaxel (MGN@DTX) were prepared and evaluated for therapeutic efficacy in nude mice bearing human prostate cancer xenografts. MGNs were prepared from PEGylated hollow gold nanospheres (HGNs) coated with folic acid and DTPTT chelate. Then, the effect of radiolabelled MGNs ((99m)Tc-MGNs) on PC-3 cell apoptosis was assessed by flow cytometry, while their binding affinity to these cells was evaluated by cell binding assays. Next, biodistribution of (99m)Tc-MGNs in xenograft bearing mice was measured by SPECT imaging. Also, DTX loading and release rates were estimated in MGN@DTX. Finally, in vitro stability in human serum and cytotoxicity of MGN@DTX were assessed, as well as their antitumor effect in xenograft bearing mice. (99m)Tc-MGNs (97.69% purity) showed good binding affinity to PC-3 cells, a specific recognition blocked by excess folic acid. Interestingly, MGN@DTX remained stable in human serum for 24 h, and exhibited higher mean cytotoxicity after NIR laser irradiation than free DTX. By day 28, tumor inhibition rates were higher in the MGN@DTX + NIR laser irradiation group compared with the DTX and MGNs + NIR laser irradiation groups. Loading chemotherapeutic drugs into MGNs can increase antitumor potency, reduce normal cell damage and decrease drug resistance, thus representing a promising approach for advanced prostate cancer treatment.
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Sodium borohydride solution, 2.0 M in triethylene glycol dimethyl ether
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Sigma-Aldrich
Sodium borohydride solution, ~12 wt. % in 14 M NaOH
Sigma-Aldrich
4-Aminothiophenol, ≥97.0%
Sigma-Aldrich
Citrate Concentrated Solution, BioUltra, for molecular biology, 1 M in H2O
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, for protein sequence analysis, ampule, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
N,N-Dimethylformamide, ACS reagent, ≥99.8%
Sigma-Aldrich
N,N-Dimethylformamide, for molecular biology, ≥99%
Sigma-Aldrich
Diethylenetriaminepentaacetic acid, ≥98% (titration)
Sigma-Aldrich
Diethylenetriaminepentaacetic acid, ≥99% (titration)
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
Sigma-Aldrich
Sodium borohydride solution, 0.5 M in 2-methoxyethyl ether
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
Sodium borohydride, 90% NaBH4 basis, pellets, diam. 8 mm, Cobalt (Doped)
Sigma-Aldrich
Sodium borohydride, powder, ≥98.0%
Sigma-Aldrich
Sodium borohydride, caplets (18 × 10 × 8 mm), 98%
Sigma-Aldrich
Sodium borohydride, purum p.a., ≥96% (gas-volumetric)
Sigma-Aldrich
Sodium borohydride, granular, 99.99% trace metals basis
Sigma-Aldrich
DCC, 1.0 M in methylene chloride
Sigma-Aldrich
N-Hydroxysuccinimide, 98%
Sigma-Aldrich
Sodium borohydride, ReagentPlus®, 99%
Sigma-Aldrich
Sodium borohydride, granular, 10-40 mesh, 98%
Sigma-Aldrich
DCC, 99%
Sigma-Aldrich
Sodium borohydride, hydrogen-storage grade, 98%