  • Search Results
  • Development of shampoo, soap and ointment formulated by green synthesised silver nanoparticles functionalised with antimicrobial plants oils in veterinary dermatology: treatment and prevention strategies.
Development of shampoo, soap and ointment formulated by green synthesised silver nanoparticles functionalised with antimicrobial plants oils in veterinary dermatology: treatment and prevention strategies.
IET nanobiotechnology (2015-08-01)
Sunita Dashrath Bansod, Manisha Subrashrao Bawaskar, Aniket Krishnarao Gade, Mahendra Kumar Rai
ABSTRACT
Many scientists have focused their research on the role of nanotechnology for the control of human pathogens, but there are also many topical pathogens present in animals, which infect animals and transfer to humans. Topical therapy is extremely important for the management of dermatological condition in animals. Therefore, the present study aims to evaluate the efficacy of biogenic silver nanoparticles (AgNPs) in combination with herbal oils against animal skin infections which may be responsible for causing infections in human beings. Here, the authors synthesised and characterised the AgNPs from Azadirachta indica. The oils were extracted from medicinal plants including Cymbopogon citratus, Cymbopogon martini, Eucalyptus globules, A. indica and Ocimum sanctum and the antifungal and antibacterial activity of plant oils along with AgNPs were evaluated. An excision wound model was used for the study of wound healing activity in rabbits. AgNPs functionalised oil has demonstrated remarkable antimicrobial activity against pathogens present on the skin of animals. The nano-functionalised antimicrobial oils were used in the formulation of shampoo, soap and ointment for veterinary dermatology. Antimicrobial products of plant origin with AgNPs are valuable, safe and have a specific role in controlling diseases. The authors believe that this approach will be a good alternative therapy to solve the continuous antibiotic resistance developed by many bacterial pathogens and will be utilised in various animal contacting areas in medicine.
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture