Novel cell-penetrating peptide targeting mitochondria.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2015-07-22)
Carmine Pasquale Cerrato, Marco Pirisinu, Efstathios Nikolaos Vlachos, Ülo Langel
ABSTRACT
Cell-penetrating peptides (CPPs) are short, nontoxic peptides with cationic and/or amphipathic properties able to cross the cellular membrane. CPPs are used for the delivery of a wide variety of cargoes, such as proteins, oligonucleotides, and therapeutic molecules. The aim of the present study was to synthesize unusually small novel CPPs targeting mitochondria based on the Szeto-Schiller peptide (SS-31) to influence intramitochondrial processes and to improve the biologic effects. All the peptides used were synthesized manually using 9-fluorenylmethyloxycarbonyl chemistry. In the first part of the study, HeLa 705, U87, and bEnd.3 cells were used as in vitro delivery model. Cells were incubated for 24 h at 37°C and 5% CO2 with different concentrations of our peptides. Cell proliferation assay was performed to evaluate cell viability. Biologic effects such as mitochondrial membrane potential and antioxidant activity were evaluated. H2O2 was used as positive control. Uptake studies were performed using peptides conjugated with 5(6)-carboxyfluorescein (FAM). Fluorescent microscopy was used to determine presence and localization of peptides into the cells. Isolated mitochondria from pretreated cells and mitochondria treated after isolation were used to confirm the targeting ability of the peptide. Uptake of FAM alone was used as negative control. Microscopy studies confirmed the ability of peptides to penetrate cell. Localization analysis showed increase in uptake by 35% compared with SS-31. Mitochondrial CPP 1 (mtCPP-1) had no effect on mitochondrial membrane potential and prevented reactive oxygen species formation in bEnd.3 cells by 2-fold compared with SS-31. No cytotoxicity was observed even at high concentration (100 µM). These data suggest that mtCPP-1 is a mitochondrial CPP and protect mitochondria from oxidative damage due to its own antioxidant activities.
Sigma-Aldrich
Water, BioPerformance Certified
Supelco
DL-Dithiothreitol solution, 1 M in H2O
SAFC
Tricine
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Tricine, BioPerformance Certified, suitable for cell culture, ≥99% (titration)
Sigma-Aldrich
Tricine, ≥99% (titration)
Sigma-Aldrich
Tricine, BioXtra, pH 4.0-6.0 (1 M in H2O), ≥99% (titration)
Sigma-Aldrich
Magnesium sulfate solution, for molecular biology, 1.00 M±0.04 M
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Diethyl ether, contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%
Sigma-Aldrich
Water, deuterium-depleted, ≤1 ppm (Deuterium oxide)
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Sigma-Aldrich
Magnesium sulfate solution, BioUltra, for molecular biology
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Diethyl ether, puriss., contains ~5 mg/L 2,6-di-tert.-butyl-4-methylphenol as stabilizer, meets analytical specification of Ph. Eur., BP, ≥99.5% (GC)
Sigma-Aldrich
Diethyl ether, ACS reagent, anhydrous, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
Diethyl ether, contains BHT as inhibitor, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
Diethyl ether, ACS reagent, ≥98.0%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor