  • Search Results
  • Coordinated Action of Aquaporins Regulates Sperm Motility in a Marine Teleost.
Coordinated Action of Aquaporins Regulates Sperm Motility in a Marine Teleost.
Biology of reproduction (2015-07-03)
Mónica Boj, François Chauvigné, Joan Cerdà
ABSTRACT
In marine teleosts, such as the gilthead seabream, several aquaporin paralogs are known to be expressed during the hyperosmotic induction of spermatozoon motility in seawater. Here, we used immunological inhibition of channel function to investigate the physiological roles of Aqp1aa, Aqp1ab, and Aqp7 during seabream sperm activation. Double immunofluorescence microscopy of SW-activated sperm showed that Aqp1aa and Aqp7 were respectively distributed along the flagellum and the head, whereas Aqp1ab accumulated in the head and in discrete areas toward the anterior tail. Inhibition of Aqp1aa reduced the rise of intracellular Ca(2+), which is independent of external Ca(2+) and normally occurs upon activation, and strongly inhibited sperm motility. Impaired Aqp1aa function also prevented the intracellular trafficking of Aqp8b to the mitochondrion, where it acts as a peroxiporin allowing H2O2 efflux and ATP production during activation. However, restoring the Ca(2+) levels with a Ca(2+) ionophore in spermatozoa with immunosuppressed Aqp1aa function fully rescued mitochondrial Aqp8b accumulation and sperm motility. In contrast, exposure of sperm to Aqp1ab and Aqp7 antibodies did not affect motility during the initial phase of activation, but latently compromised the trajectory and the pattern of movement. These data reveal the coordinated action of spatially segregated aquaporins during sperm motility activation in a marine teleost, where flagellar-localized Aqp1aa plays a dual Ca(2+)-dependent role controlling the initiation of sperm motility and the activation of mitochondrial detoxification mechanisms, while Aqp1ab and Aqp7 in the head and anterior tail direct the motion pattern.
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
2-Phenylindole, technical grade, 95%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium bicarbonate, Hybri-Max, powder, suitable for hybridoma, ≥99.5%
Sigma-Aldrich
Sodium fluoride solution
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Sodium chloride solution, 0.85%
SAFC
HEPES
SAFC
HEPES
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture