  • Search Results
  • IL-6 originated from breast cancer tissue-derived mesenchymal stromal cells may contribute to carcinogenesis.
IL-6 originated from breast cancer tissue-derived mesenchymal stromal cells may contribute to carcinogenesis.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine (2015-02-24)
Özlem Sağlam, Zehra Seda Ünal, Cansu Subaşı, Engin Ulukaya, Erdal Karaöz
ABSTRACT
Tumor microenvironment is an important factor, which sustains and promotes the tumors by inflammatory signals. Interleukin-6 (IL-6) is known as a multifunctional cytokine, which is a major activator of the signaling pathway of Janus kinases (JAKs)/signal transducer and activator of transcription 3 (STAT3). In this study, we aimed to investigate the effect of IL-6 in the tumor microenvironment on carcinogenesis. For this purpose, healthy breast tissue-derived stromal cells (HBT-SCs) and malign breast tissue-derived stromal cells (MBT-SCs) were co-cultured with MCF-7 (human breast adenocarcinoma cell line) cells using semipermeable membranes. The cell proliferation was monitored with water-soluble tetrazolium (WST) and carboxyfluorescein succinimidyl ester (CFSE) assays. Protein levels were measured by enzyme-linked immunosorbent assay (ELISA) and Western blot hybridization, while gene expressions were measured by real-time PCR. The results demonstrated that IL-6 protein levels increased significantly in the supernatants of MBT-SCs when they were co-cultured with MCF-7 cells. In accordance with this, the expression of IL-6 was significantly higher in MBT-SCs. Additionally, the expression of STAT3 in MCF-7 cells increased slightly when they were co-cultured with MBT-SCs. Considering together, there is an important interaction between tumor microenvironment and tumor cells mediated by IL-6 signaling. Thereby, the targeting on IL-6 signaling in the treatment of cancer might effectively prevent the tumor progression.
Sigma-Aldrich
Indomethacin, 98.5-100.5% (in accordance with EP)
Sigma-Aldrich
3-Amino-9-ethylcarbazole, tablet
Sigma-Aldrich
3-Amino-9-ethylcarbazole, ≥95% (HPLC), powder
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Supelco
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Methanol solution, (Methanol:Dimethyl sulfoxide 1:1 (v/v))
Sigma-Aldrich
Hematoxylin, certified by the Biological Stain Commission
Supelco
Hematoxylin
Supelco
Methanol solution, contains 0.1 % (v/v) trifluoroacetic acid, 5 % (v/v) water, suitable for HPLC
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C