  • Search Results
  • Low-aspect ratio nanopatterns on bioinert alumina influence the response and morphology of osteoblast-like cells.
Low-aspect ratio nanopatterns on bioinert alumina influence the response and morphology of osteoblast-like cells.
Biomaterials (2015-05-30)
Isabel Wittenbrink, Anne Hausmann, Karolina Schickle, Ines Lauria, Roswitha Davtalab, Morten Foss, Adrian Keller, Horst Fischer
ABSTRACT
Topographical features on the nanometer scale are known to influence cellular behavior. The response of specific cell types to various types of surface structures is currently still being investigated. Alumina ceramics play an important role as biomaterials, e.g., in medical and dental applications. In this study, we investigated the influence of nanoscale surface features with low aspect ratio (< 0.1) on the response of osteoblast-like MG-63 cells. To this end, low-energy ion irradiation was employed to produce shallow nanoscale ripple patterns on Al2O3(0001) surfaces with lateral periodicities of 24 nm and 179 nm and heights of only 0.7 and 11.5 nm, respectively. The nanopatterning was found to increase the proliferation of MG-63 cells and may lead to pseudopodia alignment along the ripples. Furthermore, focal adhesion behavior and cell morphology were analyzed. We found that MG-63 cells are able to recognize surface nanopatterns with extremely low vertical variations of less than 1 nm. In conclusion, it is shown that surface topography in the sub-nm range significantly influences the response of osteoblast-like cells.
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, anhydrous, denatured
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Sodium phosphate, 96%
Sigma-Aldrich
Hexamethyldisilazane, reagent grade, ≥99%
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
Aluminum oxide, nanopowder, <50 nm particle size (TEM)
Sigma-Aldrich
Phenylacetic acid, 99%
Sigma-Aldrich
Hexamethyldisilazane, ReagentPlus®, 99.9%
Sigma-Aldrich
N,O-Bis(trimethylsilyl)acetamide, synthesis grade, ≥95%
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Carbon, mesoporous, average pore diameter 100 Å±10 Å (typical), >99.95% trace metals basis
Sigma-Aldrich
Aluminum oxide, nanopowder, 13 nm primary particle size (TEM), 99.8% trace metals basis
Sigma-Aldrich
Carbon, mesoporous, nanopowder, <500 nm particle size (DLS), >99.95% trace metals basis
Sigma-Aldrich
Carbon nanofibers, graphitized (iron-free), composed of conical platelets, D × L 100 nm × 20-200 μm
Sigma-Aldrich
Aluminum oxide, nanoparticles, <50 nm particle size (DLS), 20 wt. % in isopropanol
Sigma-Aldrich
Carbon, mesoporous, nanopowder, graphitized, <500 nm particle size (DLS), >99.95% trace metals basis
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Phenylacetic acid, ≥99%, FCC, FG
Sigma-Aldrich
Aluminum oxide, 99.997% trace metals basis
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Aluminum oxide, nanoparticles, 30-60 nm particle size (TEM), 20 wt. % in H2O
Sigma-Aldrich
Carbon nanofibers, graphitized, platelets (conical), >98% carbon basis, D × L 100 nm × 20-200 μm
Sigma-Aldrich
Carbon, mesoporous
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.