  • Search Results
  • Inhibitory effect of Paeonia lactiflora Pallas extract (PE) on poly (I:C)-induced immune response of epidermal keratinocytes.
Inhibitory effect of Paeonia lactiflora Pallas extract (PE) on poly (I:C)-induced immune response of epidermal keratinocytes.
International journal of clinical and experimental pathology (2015-07-21)
Mi-Ra Choi, Dae-Kyoung Choi, Kyung-Cheol Sohn, Seul Ki Lim, Dong-Il Kim, Young Ho Lee, Myung Im, Young Lee, Young-Joon Seo, Chang Deok Kim, Jeung-Hoon Lee
ABSTRACT
Epidermal keratinocytes provide protective role against external stimuli by barrier formation. In addition, kertinocytes exerts their role as the defense cells via activation of innate immunity. Disturbance of keratinocyte functions is related with skin disorders. Psoriasis is a common skin disease related with inflammatory reaction in epidermal cells. We attempted to find therapeutics for psoriasis, and found that Paeonia lactiflora Pallas extract (PE) has an inhibitory potential on poly (I:C)-induced inflammation of keratinocytes. PE significantly inhibited poly (I:C)-induced expression of crucial psoriatic cytokines, such as IL-6, IL-8, CCL20 and TNF-α, via down-regulation of NF-κB signaling pathway in human keratinocytes. In addition, PE significantly inhibited poly (I:C)-induced inflammasome activation, in terms of IL-1β and caspase-1 secretion. Finally, PE markedly inhibited poly (I:C)-increased NLRP3, an important component of inflammasome. These results indicate that PE has an inhibitory effect on poly (I:C)-induced inflammatory reaction of keratinocytes, suggesting that PE can be developed for the treatment of psoriasis.
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
hEGF, EGF, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
EGF human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
Epidermal Growth Factor, human, animal component free, EGF, recombinant, expressed in Escherichia coli, >97% (SDS-PAGE)