  • Search Results
  • Effects of Amoxicillin and Clavulanic Acid on the Spontaneous Mechanical Activity of Juvenile Rat Duodenum.
Effects of Amoxicillin and Clavulanic Acid on the Spontaneous Mechanical Activity of Juvenile Rat Duodenum.
Journal of pediatric gastroenterology and nutrition (2015-04-07)
Steven L Ciciora, Kent C Williams, Cheryl E Gariepy
ABSTRACT
There are a limited number of medications for the treatment of foregut dysmotility. Enteral amoxicillin/clavulanic acid induces phase III duodenal contractions in a fasting pediatric patient. The mechanism by which this occurs is unknown. We examined the individual contributions of amoxicillin and clavulanic acid on the spontaneous mechanical activity of juvenile rat duodenum to better understand this phenomenon. Duodenal segments from juvenile rats were longitudinally attached to force transducers in organ baths. Samples were cumulatively exposed to amoxicillin or clavulanic acid. Separate samples were exposed to carbachol alone to assess response in both the presence and absence of amoxicillin or clavulanic acid. Basal tone, frequency, and amplitude of contractions were digitized and recorded. The amplitude of the spontaneous contractions increased with amoxicillin. Inhibition of neuronal activity prevented this effect. Clavulanic acid did not affect the spontaneous contractions. Basal tone and the rate of contractions did not differ with either drug. Stimulation with carbachol in the presence of amoxicillin caused a statistically significant increase in the contractility compared with carbachol alone. Amoxicillin alters the spontaneous longitudinal mechanical activity of juvenile rat duodenum. Our results suggest that amoxicillin modulates the spontaneous pattern of cyclic mechanical activity of duodenal smooth muscle through noncholinergic, neurally mediated mechanisms. Our work provides an initial physiologic basis for the therapeutic use of amoxicillin in patients with gastrointestinal dysmotility.
Sigma-Aldrich
Magnesium sulfate, puriss. p.a., drying agent, anhydrous, ≥98.0% (KT), powder (very fine)
Sigma-Aldrich
Magnesium sulfate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Magnesium sulfate, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥97%
Sigma-Aldrich
Magnesium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Amoxicillin, 95.0-102.0% anhydrous basis
Sigma-Aldrich
Magnesium sulfate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Magnesium sulfate, anhydrous, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Magnesium sulfate, anhydrous, reagent grade, ≥97%
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Magnesium sulfate solution, for molecular biology, 1.00 M±0.04 M
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Carbamoylcholine chloride, ≥98% (titration), crystalline
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Atropine, ≥99% (TLC), powder
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Magnesium sulfate solution, BioUltra, for molecular biology
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
SAFC
Sodium chloride solution, 5 M