MilliporeSigma
  • Effect of addition of thermally modified cowpea protein on sensory acceptability and textural properties of wheat bread and sponge cake.
Effect of addition of thermally modified cowpea protein on sensory acceptability and textural properties of wheat bread and sponge cake.
Food chemistry (2015-10-17)
Lydia Campbell, Stephen R Euston, Mohamed A Ahmed
ABSTRACT
This paper investigates the sensory acceptability and textural properties of leavened wheat bread and sponge cake fortified with cow protein isolates that had been denatured and glycated by thermal treatment. Defatted cowpea flour was prepared from cow pea beans and the protein isolate was prepared (CPI) and thermally denatured (DCPI). To prepare glycated cowpea protein isolate (GCPI) the cowpea flour slurry was heat treated before isolation of the protein. CPI was more susceptible to thermal denaturation than GCPI as determined by turbidity and sulphydryl groups resulting in greater loss of solubility. This is attributed to the higher glycation degree and higher carbohydrate content of GCPI as demonstrated by glycoprotein staining of SDS PAGE gels. Water absorption of bread dough was significantly enhanced by DCPI and to a larger extent GCPI compared to the control, resulting in softer texture. CPI resulted in significantly increased crumb hardness in baked bread than the control whereas DCPI or GCPI resulted in significantly softer crumb. Bread fortified with 4% DCPI or GCPI was similar to control as regards sensory and textural properties whereas 4% CPI was significantly different, limiting its inclusion level to 2%. There was a trend for higher sensory acceptability scores for GCPI containing bread compared DCPI. Whole egg was replaced by 20% by GCPI (3.5%) in sponge cake without affecting the sensory acceptability, whereas CPI and DCPI supplemented cakes were significantly different than the control.
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium acetate solution, BioUltra, for molecular biology, ~3 M in H2O
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, for luminescence, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90%
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FCC, FG
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium acetate, anhydrous, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium acetate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium acetate, puriss. p.a., ACS reagent, reag. Ph. Eur., anhydrous
Sigma-Aldrich
Sodium acetate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium acetate, anhydrous, for molecular biology, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)