  • Search Results
  • Thermosensitive chitosan-based hydrogels for sustained release of ferulic acid on corneal wound healing.
Thermosensitive chitosan-based hydrogels for sustained release of ferulic acid on corneal wound healing.
Carbohydrate polymers (2015-10-11)
Ching-Yao Tsai, Lin-Chung Woung, Jiin-Cherng Yen, Po-Chen Tseng, Shih-Hwa Chiou, Yen-Jen Sung, Kuan-Ting Liu, Yung-Hsin Cheng
ABSTRACT
Oxidative damage to cornea can be induced by alkaline chemical burn which may cause vision loss or blindness. Recent studies showed that exogenous application of natural antioxidants may be a potential treatment for corneal wound healing. However, low ocular bioavailability and short residence time are the limiting factors of topically administered antioxidants. Ferulic acid (FA) is a natural phenolic compound and an excellent antioxidant. The study was aimed to investigate the effects of FA in corneal epithelial cells (CECs) under oxidative stress and evaluate the feasibility of use the thermosensitive chitosan-based hydrogel containing FA for corneal wound healing. The results demonstrated that post-treatment of FA on CECs could decrease the inflammation-level and apoptosis. In the rabbit corneal alkali burn model, post-treatment FA-loaded hydrogel may promote the corneal wound healing. The results of study suggest that FA-loaded hydrogel may have the potential applications in treating corneal alkali burn.
Sigma-Aldrich
Luminol, 97%
Sigma-Aldrich
Water-16O, ≥99.94 atom % 16O
Sigma-Aldrich
Gelatin from porcine skin, gel strength 300, Type A
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Luminol, ≥97% (HPLC)
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, HPLC/spectrophotometric grade
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O