  • Search Results
  • Autophagy contributes to the chemo-resistance of non-small cell lung cancer in hypoxic conditions.
Autophagy contributes to the chemo-resistance of non-small cell lung cancer in hypoxic conditions.
Respiratory research (2015-11-11)
Jin Gu Lee, Ju Hye Shin, Hyo Sup Shim, Chang Young Lee, Dae Joon Kim, Young Sam Kim, Kyung Young Chung
ABSTRACT
The development of chemo-resistance in non-small lung cancer is a major obstacle in treating patients. Hypoxia is a commonly faced microenvironment in solid tumor and suggested to be related to both autophagy and chemo-resistance. In this study, we investigated the role of hypoxia-induced autophagy in acquiring chemo-resistance in both cancer cell (A549) and human cancer tissue Hypoxic exposure (1 % O2) of A549 cell stimulated autophagic induction in cancer cells, shown by increase of LC3BI to LC3BII conversion and decrease of p62/sequestosome1 in Western blot, increased GFP-LC puncta in confocal microscopy, and increased number of double-membrane autophagic vacuoles in electron micrographs. Hypoxic exposure also induced resistance of cancer cells to cisplatin, and LC3B siRNA restored the sensitivity of cancer cells to chemotherapy. Furthermore, Human lung cancer tissues that experienced chemotherapy showed increase of LC3BI to LC3BII conversion and decrease of p62/sequestosome1 compared with chemo-naïve cancer tissue in Western blot. Autophagy may play an important role in acquiring resistance to chemotherapy in lung cancer and hypoxia related pathway seems to be involved in autophagy induction.
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline
Sigma-Aldrich
trans-Platinum(II)diammine dichloride
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, granular
Sigma-Aldrich
Sodium sulfate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Sodium sulfate, puriss., meets analytical specification of Ph. Eur., BP, USP, anhydrous, 99.0-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium sulfate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium sulfate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium sulfate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., anhydrous, ≥99.0%
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, powder
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Supelco
Meprobamate solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture