  • Search Results
  • Synthesis, structural studies, kinetic stability, and oxidation catalysis of the late first row transition metal complexes of 4,10-dimethyl-1,4,7,10-tetraazabicyclo[6.5.2]pentadecane.
Synthesis, structural studies, kinetic stability, and oxidation catalysis of the late first row transition metal complexes of 4,10-dimethyl-1,4,7,10-tetraazabicyclo[6.5.2]pentadecane.
Dalton transactions (Cambridge, England : 2003) (2015-04-16)
Dallas L Matz, Donald G Jones, Kimberly D Roewe, Michael-Joseph Gorbet, Zhan Zhang, Zhuqi Chen, Timothy J Prior, Stephen J Archibald, Guochuan Yin, Timothy J Hubin
ABSTRACT
Synthetic details for 4,11-dimethyl-1,4,8,11-tetraazabicyclo[6.5.2]pentadecane, the dimethyl ethylene cross-bridged homocyclen ligand are presented for the first time. Its novel Mn(2+), Fe(2+), Mn(3+), and Fe(3+) complexes have been synthesized and characterized. X-ray crystal structures were obtained for both manganese complexes, along with five additional Co(3+), Cu(2+), and Zn(2+) structures, the first structural characterization of complexes of this ligand. Each complex has the cis-V configuration of the cross-bridged macrocycle ring, leaving cis labile binding sites for interaction of the complex with oxidants and/or substrates. The copper(II) complex kinetic stability in 5 M HCl and at elevated temperatures was determined and compared to related complexes in the literature. The electronic properties of the manganese and iron complexes were evaluated using solid state magnetic moment determination and acetonitrile solution electronic spectroscopy, revealing high spin metal complexes in all cases. Cyclic voltammetry in acetonitrile of the divalent iron and manganese complexes revealed reversible redox processes, suggesting catalytic reactivity involving electron transfer processes are possible for both complexes. Screening of the Mn(2+) and Fe(2+) complexes for oxidation catalysis using hydrogen peroxide as the terminal oxidant showed both complexes are worthy of continued development.
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Iodomethane, purum, ≥99.0% (GC)
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
Sodium borohydride solution, 0.5 M in 2-methoxyethyl ether
Sigma-Aldrich
Iodomethane, contains copper as stabilizer, ReagentPlus®, 99%
Sigma-Aldrich
Sodium borohydride solution, ~12 wt. % in 14 M NaOH
Sigma-Aldrich
Iodomethane, contains copper as stabilizer, ReagentPlus®, 99.5%
Sigma-Aldrich
Sodium borohydride solution, 2.0 M in triethylene glycol dimethyl ether
Sigma-Aldrich
Iodomethane-12C, ≥99.9 atom % 12C, ≥99% (CP), contains copper as stabilizer
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Sodium borohydride, 90% NaBH4 basis, pellets, diam. 8 mm, Cobalt (Doped)
Sigma-Aldrich
Sodium borohydride, powder, ≥98.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Sodium borohydride, caplets (18 × 10 × 8 mm), 98%
Sigma-Aldrich
Sodium borohydride, purum p.a., ≥96% (gas-volumetric)
Sigma-Aldrich
Sodium borohydride, granular, 99.99% trace metals basis
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethanol, anhydrous, denatured
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium borohydride, ReagentPlus®, 99%
Sigma-Aldrich
Sodium borohydride, granular, 10-40 mesh, 98%
Sigma-Aldrich
Iodomethane solution, 2.0 M in tert-butyl methyl ether, contains copper as stabilizer
Sigma-Aldrich
Sodium borohydride, hydrogen-storage grade, 98%