  • Search Results
  • Enhanced Control of Bladder-Associated Tumors Using Shrimp Anti-Lipopolysaccharide Factor (SALF) Antimicrobial Peptide as a Cancer Vaccine Adjuvant in Mice.
Enhanced Control of Bladder-Associated Tumors Using Shrimp Anti-Lipopolysaccharide Factor (SALF) Antimicrobial Peptide as a Cancer Vaccine Adjuvant in Mice.
Marine drugs (2015-05-27)
Han-Ning Huang, Venugopal Rajanbabu, Chieh-Yu Pan, Yi-Lin Chan, Jyh-Yih Chen, Chang-Jer Wu
ABSTRACT
Shrimp anti-lipopolysaccharide factor (SALF) is an antimicrobial peptide with reported anticancer activities, such as suppression of tumor progression. In this study, we prepared a potential cancer vaccine comprised of SALF in conjunction with the cell lysate of inactivated murine bladder carcinoma cells (MBT-2), and evaluated its efficacy in a mouse tumor model. Our study shows that SALF added to cell culture media inhibits growth progression of MBT-2, and that SALF together with inactivated MBT-2 lysate elevates the level of inflammasome activity, and modulates the levels of IL-1β, MCP-1, IL-6, IL-12, and TNF-α in mouse macrophages. Immunization of 7, 14, and 21 day-old mice with the vaccine prevented growth of MBT-2 cell-mediated tumors. The vaccine was found to enhance expression of T-cell, cytotoxic T cells, and NK cells in the immunized mice groups. Recruitment of macrophages, T-helper cells, and NK cells was enhanced, but levels of VEGF were decreased in immunized mice. This report provides empirical evidence that our SALF as vaccine adjuvant enhances antitumor immunity in mice.
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline
Sigma-Aldrich
trans-Platinum(II)diammine dichloride
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Tris(tert-butoxy)silanol, packaged for use in deposition systems
Sigma-Aldrich
Diethyl azodicarboxylate solution, 40 wt. % in toluene
Sigma-Aldrich
Tris(tert-butoxy)silanol, 99.999%