  • Search Results
  • The AKT signaling pathway sustains the osteogenic differentiation in human dental follicle cells.
The AKT signaling pathway sustains the osteogenic differentiation in human dental follicle cells.
Molecular and cellular biochemistry (2015-05-20)
S Viale-Bouroncle, C Klingelhöffer, T Ettl, C Morsczeck
ABSTRACT
Signaling transduction pathways are established by interactions between growth factors, protein kinases, and transcription factors, and they play a crucial role in tooth development. Precursor cells of the dental follicle (DFCs) are used for in vitro studies about molecular mechanisms during periodontal development. Previous studies have already shown that the growth factor BMP2 and the transcription factor EGR1 are involved in the osteogenic differentiation in DFCs while interactions with protein kinase-based pathways remain elusive. In this current study, we investigated the role of the AKT kinase signaling pathway for the osteogenic differentiation in DFCs. The AKT signaling pathway was activated in DFCs after the induction of the osteogenic differentiation by BMP2. The inhibition of AKT in DFCs repressed the differentiation and the expression of the transcription factor EGR1. Interestingly, EGR1 bound to the phosphorylated form of SMAD1/5 (pSMAD). The binding of pSMAD to EGR1 was increased after the induction with BMP2. Moreover, the overexpression EGR1 increased the osteogenic differentiation of DFCs. Our results suggest that the AKT signaling pathway submits the BMP2-dependent osteogenic differentiation in DFCs via the expression of the transcription factor EGR1.
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Lithium dodecyl sulfate, BioReagent, for molecular biology, suitable for electrophoresis
Sigma-Aldrich
Lithium dodecyl sulfate, ≥98.5% (GC)
Sigma-Aldrich
Lithium dodecyl sulfate, BioXtra, ≥98.5% (GC)
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
Dexamethasone 21-phosphate disodium salt, ≥98%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)