MilliporeSigma
Cytotoxicity of commonly used luting cements -An in vitro study.
Dental materials journal (2015-04-24)
Rita Trumpaite-Vanagiene, Virginija Bukelskiene, Jolanta Aleksejuniene, Alina Puriene, Daiva Baltriukiene, Vygandas Rutkunas
ABSTRACT
The study aimed to 1) evaluate the cytotoxicity of luting cements: Hoffmann's Zinc Phosphate (Hoffmann's ZP), GC Fuji Plus Resin Modified Glass Ionomer (Fuji Plus RMGI) and 3M ESPE RelyX Unicem Resin Cement (RelyX Unicem RC) and 2) test if pre-washing reduces the cements' cytotoxicity. In vitro human gingival fibroblast (HGF) culture model was chosen. The cytotoxicity was evaluated by MTT test, the cell viability -by staining the cells with AO/EB dye mixture. The means±SD of Cell Survival Ratio (CSR%) were compared among different cement types under two testing conditions, with or without cement pre-washing. The CSR%s were compared by ANOVA and linear multiple regression (LMR). Hoffmann's ZPC was less cytotoxic, while Fuji Plus RMGIC and RelyX Unicem RC were more cytotoxic (ANOVA, p<0.001). The type of cement and cement pre-washing jointly explained 90% of cell survival (LMR, p<0.001, adjusted squared R=0.889). The commonly used luting cements such as Hoffmann's ZP, Fuji Plus RMGI and RelyX Unicem RC may have a cytotoxic potential.
Sigma-Aldrich
Ethidium bromide, BioReagent, for molecular biology, powder
Sigma-Aldrich
Ethidium bromide, ~95% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Ethidium bromide solution, BioReagent, for molecular biology, 10 mg/mL in H2O
Sigma-Aldrich
Ethidium bromide solution, BioReagent, for molecular biology, 500 μg/mL in H2O
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Supelco
Pyridaben, PESTANAL®, analytical standard
Sigma-Aldrich
Ethidium bromide solution, for fluorescence, ~1% in H2O
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%