  • Search Results
  • Effect of Ca(OH)2 pretreatment on extruded rice straw anaerobic digestion.
Effect of Ca(OH)2 pretreatment on extruded rice straw anaerobic digestion.
Bioresource technology (2015-08-02)
Yu Gu, Yalei Zhang, Xuefei Zhou
ABSTRACT
It has been proven that extrusion can change the structure of rice straw and increase biogas production, but the effect of a single pretreatment is limited. Ca(OH)2 pretreatment was used to enhance the enzyme hydrolysis and biogas production of extruded rice straw. After Ca(OH)2 pretreatment, the glucose and xylose conversion rates in enzymatic hydrolysis increased from 36.0% and 22.4% to 66.8% and 50.2%, respectively. The highest biogas production observed in 8% and 10% Ca(OH)2 pretreated rice straw reached 564.7mL/g VS and 574.5mL/g VS, respectively, which are 34.3% and 36.7% higher than the non-Ca(OH)2-loaded sample. The Ca(OH)2 pretreatment can effectively remove the lignin and increase the fermentable sugar content. The structural changes in the extruded rice straw have also been analyzed by XRD, FTIR, and SEM. Considering all of the results, an 8% Ca(OH)2 loading rate is the best option for the pretreatment of extruded rice straw.
Product Description
Carbon - Vitreous, foil, 50x50mm, thickness 1.0mm, glassy carbon
Carbon - Vitreous, rod, 100mm, diameter 7.0mm, glassy carbon
Carbon - Vitreous, rod, 200mm, diameter 3.0mm, glassy carbon
Carbon - Vitreous, foil, 100x100mm, thickness 2.0mm, glassy carbon
Carbon - Vitreous, foil, 8x8mm, thickness 0.5mm, glassy carbon
Carbon - Vitreous, foam, 150x150mm, thickness 2.5mm, bulk density 0.05g/cm3, porosity 96.5%
Carbon - Vitreous, foam, 275x330mm, thickness 3.2mm, bulk density 0.05g/cm3, porosity 96.5%
Carbon - Vitreous, rod, 200mm, diameter 10mm, glassy carbon
Carbon - Vitreous, foil, 10x10mm, thickness 1.0mm, glassy carbon
Carbon - Vitreous, foam, 150x150mm, 0.05g.cmué, porosity 96.5%, 24 pores/cm
Carbon - Vitreous, foil, 10x10mm, thickness 4.0mm, glassy carbon
Carbon - Vitreous, foil, 100x100mm, thickness 1.0mm, glassy carbon
Carbon - Vitreous, rod, 200mm, diameter 5.0mm, glassy carbon
Carbon - Vitreous, rod, 100mm, diameter 1.0mm, glassy carbon
Carbon - Vitreous, rod, 5mm, diameter 3.0mm, glassy carbon
Carbon - Vitreous, foam, 300x300mm, thickness 30mm, bulk density 0.05g/cm3, porosity 96.5%
Carbon - Vitreous, foil, 50x50mm, thickness 4.0mm, glassy carbon
Carbon - Vitreous, foam, 150x150mm, thickness 3.2mm, bulk density 0.05g/cm3, porosity 96.5%
Carbon - Vitreous, rod, 50mm, diameter 1.0mm, glassy carbon
Carbon - Vitreous, foam, 300x300mm, thickness 20mm, bulk density 0.05g/cm3, porosity 96.5%
Carbon - Vitreous, foam, 275x330mm, 0.05g.cmué, porosity 96.5%, 24 pores/cm
Carbon - Vitreous, rod, 200mm, diameter 1.0mm, glassy carbon
Carbon - Vitreous, foil, 10x10mm, thickness 2.0mm, glassy carbon
Carbon - Vitreous, foil, 25x25mm, thickness 4.0mm, glassy carbon
Carbon - Vitreous, foil, 25x25mm, thickness 0.5mm, glassy carbon
Carbon - Vitreous, foil, 100x100mm, thickness 6.0mm, glassy carbon
Carbon - Vitreous, foil, 8mm disks, thickness 1.0mm, glassy carbon
Carbon - Vitreous, rod, 100mm, diameter 5.0mm, glassy carbon
Carbon - Vitreous, rod, 100mm, diameter 3.0mm, glassy carbon
Carbon - Vitreous, foil, 10mm disks, thickness 0.2mm, glassy carbon