  • Search Results
  • Effect of bioreduced graphene oxide on anaerobic biotransformation of nitrobenzene in an anaerobic reactor.
Effect of bioreduced graphene oxide on anaerobic biotransformation of nitrobenzene in an anaerobic reactor.
Environmental technology (2015-06-27)
Jing Wang, Haikun Zhang, Di Wang, Hong Lu, Jiti Zhou
ABSTRACT
Bioreduced graphene oxide (BRGO) has been proven to be capable of accelerating nitrobenzene (NB) biotransformation by anaerobic sludge (AS). To realize its application, in the present study, two continuous anaerobic reactors (R1 with BRGO/AS composite; R2 with AS) were employed to treat NB-containing wastewater during a long-term run. Compared with R2, the start-up time of R1 was shortened from 70 to 45 d and R1 exhibited better removal efficiency (87% of R1 and 74% of R2 with an influent NB concentration of 200 mg L(-1) at hydraulic retention time = 24 h, NaCl = 3%). Moreover, R1 exhibited better stability with over 81% NB removal efficiency within 90 d. Further study demonstrated that the presence of BRGO facilitated microorganisms to secrete extracellular polymeric substances, resulting in the higher electrochemical and dehydrogenase activities of R1 compared with those of R2.
Sigma-Aldrich
(−)-Riboflavin, meets USP testing specifications
Sigma-Aldrich
(−)-Riboflavin, from Eremothecium ashbyii, ≥98%
Sigma-Aldrich
(−)-Riboflavin, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98%
Sigma-Aldrich
Titanium tetrachloride, packaged for use in deposition systems
Sigma-Aldrich
Titanium(IV) chloride solution, 1.0 M in toluene
Sigma-Aldrich
Titanium(IV) chloride solution, 1.0 M in methylene chloride
Sigma-Aldrich
Titanium(IV) chloride solution, 0.09 M in 20% HCl
Supelco
Riboflavin (B2), analytical standard
Sigma-Aldrich
Titanium(IV) chloride, ReagentPlus®, 99.9% trace metals basis
Sigma-Aldrich
Titanium(IV) chloride, ≥99.995% trace metals basis