  • Search Results
  • The resveratrol oligomers, cis- and trans-gnetin H, from Paeonia suffruticosa seeds inhibit the growth of several human cancer cell lines.
The resveratrol oligomers, cis- and trans-gnetin H, from Paeonia suffruticosa seeds inhibit the growth of several human cancer cell lines.
Journal of ethnopharmacology (2015-04-12)
Ying Gao, Chunnian He, Ran Ran, Dabing Zhang, Dawei Li, Pei-Gen Xiao, Elliot Altman
ABSTRACT
Paeonia suffruticosa Andrews (PSE) is a well-known Chinese medicine that has been widely used as an anti-tumor, anti-oxidative and anti-inflammatory agent. cis- and trans-gnetin H are two resveratrol oligomers isolated from the seeds of PSE. Although resveratrol is widely considered to be one of the most valuable natural chemopreventive agents and there are numerous studies on the antitumor activities of resveratrol, little is known about the antitumor properties of cis- and trans-gnetin H. The inhibitory effects of cis- and trans-gnetin H in different human cancer cell lines were assessed using fluorescent viability tests. Cytotoxicity in human lung and breast cancer cells was detected via nuclear condensation, cell permeability, and changes in the mitochondrial membrane potential (∆ψm). Apoptosis in human lung and breast cancer cells was assessed by flow cytometry, a luminescence assay and high-content screening analysis. Finally, a xenograft mice model was used to examine the efficacy of cis-gnetin H on lung tumors. cis- and trans-gnetin H have superior activity in inhibiting the proliferation of four human cancer cell lines, A549 (lung), BT20 (breast), MCF-7 (breast) and U2OS (osteosarcoma), and promote cell apoptosis, while having a minimal effect on two normal human epithelial cell lines, HPL1A (lung) and HMEC (breast) used as controls. cis- and trans-gnetin H promote apoptosis by releasing mitochondria cytochrome c, activating caspase 3/7 and inhibiting NF-κB activation. Flow cytometry analysis shows that cis- or trans-gnetin H arrested the cell cycle of cancer cells at the G0-G1 phase. Moreover, cis-gnetin H suppressed the growth of xenograft lung tumors in mice. Collectively, our findings demonstrate the promise of the natural compounds cis- and trans-gnetin H as candidates for cancer chemotherapy agents.
Sigma-Aldrich
Valinomycin, ≥98% (TLC), ≥90% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Propidium iodide solution, solution (1.0 mg/ml in water)
Sigma-Aldrich
Valinomycin, ≥99.0% (TLC)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Valinomycin, Ready Made Solution, ~1 mg/mL in DMSO
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)