  • Search Results
  • Biodegradation of di-n-Butyl Phthalate by Achromobacter sp. Isolated from Rural Domestic Wastewater.
Biodegradation of di-n-Butyl Phthalate by Achromobacter sp. Isolated from Rural Domestic Wastewater.
International journal of environmental research and public health (2015-10-31)
Decai Jin, Xiao Kong, Yujie Li, Zhihui Bai, Guoqiang Zhuang, Xuliang Zhuang, Ye Deng
ABSTRACT
A bacterial strain W-1, isolated from rural domestic wastewater, can utilize the environmental hormone di-n-butyl phthalate (DBP) as the sole carbon and energy source. The isolated bacterium species was confirmed to belong to the genus Achromobacter based on its 16S rRNA gene sequence. The results of substrate utilization tests showed that the strain W-1 could utilize other common phthalates and phenol. High-performance liquid chromatography analysis revealed that the optimal conditions for DBP degradation were pH 7.0, 35 °C, and an agitation rate of 175 rpm. Under these conditions, 500 mg/L of DBP was completely degraded within 30 h. The effects of heavy metals (50 mg/L Cu(2+) and 500 mg/L Pb(2+)) and surfactants (100 mg/L SDS and 500 mg/L Tween 20) on DBP degradation were investigated. The results demonstrated that Cu(2+) and SDS severely inhibited DBP degradation and Pb(2+) weakly inhibited DBP degradation, while Tween 20 greatly enhanced DBP degradation. Furthermore, phthalate degradation genes were found to be located on a plasmid present in Achromobacter sp. W-1.
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Dess-Martin periodinane, 97%
Sigma-Aldrich
Dimethyl phthalate, ≥99%
Sigma-Aldrich
Dibutyl phthalate, 99%
Sigma-Aldrich
Dimethyl phthalate, ≥99%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, meets analytical specification of Ph. Eur., BP, E339, 98.5-102.5% (T)
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, puriss. p.a., crystallized, ≥99.0% (T)
Sigma-Aldrich
Hexane, Laboratory Reagent, ≥95%
Sigma-Aldrich
Diethyl phthalate, 99.5%
Sigma-Aldrich
Diethyl pyrocarbonate, 96% (NT)
Supelco
Ethyl acetate, HPLC grade, 99.8%
Sigma-Aldrich
Diethyl phthalate, ≥99%
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Diethyl pyrocarbonate, 96% (NT)
Sigma-Aldrich
Ethyl acetate