  • Search Results
  • The cardioprotective effect of hypoxic and ischemic preconditioning in dogs with myocardial ischemia-reperfusion injury using a double-bypass model.
The cardioprotective effect of hypoxic and ischemic preconditioning in dogs with myocardial ischemia-reperfusion injury using a double-bypass model.
Life sciences (2015-09-15)
Chun-Ling Chen, Hong Zheng, Yan Xuan, Ablikim Amat, Lin Chen, Jin Yu, Jiang Wang
ABSTRACT
The effects of preconditioning on cardioprotection have mainly been studied in vitro. No sufficient in vivo experiments have been performed to optimize ischemic preconditioning (IPC) or hypoxic preconditioning (HPC) for clinical applications. The purpose of this study was to establish a canine double-bypass model to examine the effect of IPC and HPC on cardiomyocytes and heart function. A double-bypass procedure to enable independent control of systemic and coronary circulation was established in dogs. The animals were divided into control, HPC, and IPC groups (n=6 each). Indicators of cardiac function, including cardiodynamics, hemodynamics, ATP, and cardiac troponin I (cTnI) levels; myocardium morphology; and myocardiocyte apoptosis were determined. Both IPC and HPC attenuated the reperfusion-induced decrease in left ventricular end systolic pressure seen in the control group. Both the HPC and IPC groups had lower serum cTnI levels, better myocardiocyte histology, and lower rates of apoptosis compared to the control group without preconditioning. HPC reduced the abnormal cardiomyocyte histology and apoptosis to a greater extent than IPC, and only HPC significantly restored the depletion of ATP. This study demonstrates the effectiveness of the double-bypass model for the optimized study of both HPC and IPC. The results suggest that HPC may provide better cardioprotection than IPC.
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Sigma-Aldrich
D.E.R. 332, used as embedding medium
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Supelco
Acetone solution, certified reference material, 2000 μg/mL in methanol: water (9:1)
Sigma-Aldrich
Nitrogen, ≥99.998%