  • Search Results
  • c-ETS transcription factors play an essential role in the licensing of human MCM4 origin of replication.
c-ETS transcription factors play an essential role in the licensing of human MCM4 origin of replication.
Biochimica et biophysica acta (2015-09-15)
Kaveri Sidhu, Vijay Kumar
ABSTRACT
In metazoans, DNA replication is a highly regulated and ordered process that occurs during the S phase of cell cycle. It begins with the licensing of origins of replication usually found in close proximity of actively transcribing genes owing perhaps to a profound influence of transcription factors on the epigenetic signatures and architecture of chromatin. Here we show that ETS transcription factors are novel regulators of MCM4 origin, whose binding sites are localized between two divergently transcribing MCM4 and PRKDC genes. c-ETS1 and c-ETS2 were recruited to the MCM4 origin respectively during the S and G1 phases of cell cycle. c-ETS2 binding was facilitated by an active chromatin distinguished by acetylated histone H3 orchestrated by histone acetyl transferase GCN5 and followed by HBO1 mediated histone H4 acetylation. Interestingly, c-ETS2 overexpression led to increased BrdU incorporation in the S phase cells while its down-regulation by RNA interference compromised the loading of pre-replicative complex at the origin. Conversely, the recruitment of c-ETS1 at the origin coincided with histone H3 methylation signature characteristic of closed chromatin conformation. As expected, enforced expression of c-ETS1 severely compromised DNA replication whereas its down-regulation enhanced DNA replication as evident from increased BrdU incorporation. Thus, c-ETS transcription factors appear to be key regulators of MCM4 origin where c-ETS2 seems to promote DNA replication whereas c-ETS1 functions as a negative regulator.
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, HPLC/spectrophotometric grade
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology
Sigma-Aldrich
Ethanol, puriss. p.a., absolute, ≥99.8% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydroxyurea, 98%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Methyl methanesulfonate, 99%
Sigma-Aldrich
Phenol Red, ACS reagent
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Phenol Red, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
MISSION® esiRNA, targeting human ETS1
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Sodium pyrophosphate tetrabasic, ≥95%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 5 M
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
Thymidine, ≥99%