  • Search Results
  • New analytical method for the study of metabolism of swertiamarin in rats after oral administration by UPLC-TOF-MS following DNPH derivatization.
New analytical method for the study of metabolism of swertiamarin in rats after oral administration by UPLC-TOF-MS following DNPH derivatization.
Biomedical chromatography : BMC (2015-01-23)
Xiuhong Wu, Shuhan Tang, Yan Jin, Yan Zhang, Masao Hattori, Xijun Wang, Hailong Zhang, Zhigang Wang
ABSTRACT
The metabolism of swertiamarin in vivo was studied by LC-MS following 2,4-dinitrophenylhydrazine derivatization. The ionization efficiency of the main metabolite erythrocentaurin was greatly enhanced by the new analytical method developed, and erythrocentaurin was successfully detected for the first time in rat plasma after oral administration of swertiamarin. Methyl 4-formylbenzoate was used as the internal standard to quantify erythrocentaurin in rat plasma in negative mode by UPLC-TOF-MS, and it was found that erythrocentaurin reached the maximum mean plasma concentration of 425.8 ± 127.6 ng/mL at about 2 h after oral administration of swertiamarin at a dose of 200 mg/kg. A metabolic pathway of swertiamarin to erythrocentaurin was proposed. Swertiamarin is first hydrolyzed by bacterial β-glucusidase to give the aglycone, which is readily converted to erythrocentaurin. The monoterpene compound swertiamarin was found to be metabolized to dihydroisocoumarin and alkaloid compounds in vivo, which may be responsible for the pharmacological effect of swertiamarin. The results may shed light on the clinical efficacy of swertiamarin and the new analytical method may assist in studies for the metabolism of other natural iridoids and secoiridoids in vivo.
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 40.0% 2-propanol, 0.05% formic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Methyl 4-formylbenzoate, 99%
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 0.1 % (v/v) formic acid, 5 % (v/v) water, suitable for HPLC
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
2,4-Dinitrophenylhydrazine, reagent grade, 97%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile