  • Search Results
  • Pre- or post-ischemic bilirubin ditaurate treatment reduces oxidative tissue damage and improves cardiac function.
Pre- or post-ischemic bilirubin ditaurate treatment reduces oxidative tissue damage and improves cardiac function.
International journal of cardiology (2015-09-21)
Bhavisha Bakrania, Eugene F Du Toit, Karl-Heinz Wagner, John P Headrick, Andrew C Bulmer
ABSTRACT
Unconjugated bilirubin (UCB), an endogenous antioxidant, may protect the heart against ischemia-reperfusion (I-R) injury. However, the 'cardioprotective' potential of bilirubin therapy remains unclear. We tested whether pre- or post-ischemic treatment of ex vivo perfused hearts with bilirubin ditaurate (BRT) improves post-ischemic functional outcomes and myocardial oxidative damage. Isolated Langendorff perfused hearts (male, Wistar rats) were treated with 50 μM BRT for 30 min before (Pre) or after (Post) 30 min of zero-flow ischemia. Functional outcomes were monitored, with myocardial damage estimated from creatine kinase efflux, infarct size, and left ventricular lipid/protein oxidation assessed by measuring malondialdehyde and protein carbonyls. Ischemia induced contractile dysfunction and cellular injury, with both BRT treatments improving I-R outcomes. Final post-ischemic recoveries for left ventricular diastolic/developed pressures were significantly enhanced in treated groups: end-diastolic pressure (Control, 78±14, Pre, 51±15*, Post, 51±13 mm Hg*); left ventricular developed pressure, (LVDP; Control 44±15, Pre, 71±19*, Post, 84±13 mm Hg*). Myocardial injury/infarction (MI) was also significantly reduced with BRT treatment: post-ischemic creatine kinase efflux (Control, 1.24±0.41, Pre, 0.86±0.31*, Post, 0.51±0.29 U/g/mL*; infarct size, Control, 67±17, Pre, 39±15*, Post, 22±11%*). These changes were accompanied by significantly reduced malondialdehyde and protein carbonyl content in Pre and Post treated hearts (*P<0.05 vs. Control). These data collectively reveal significant cardioprotection upon BRT treatment, with post-treatment being particularly effective. Significant reductions in infarct size and lipid and protein oxidation indicate a mechanism related to protection from oxidative damage and indicate the potential utility of this molecule as a post-MI treatment.
Sigma-Aldrich
Phosphoric acid, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, 85.0-88.0%
Sigma-Aldrich
Titanium(IV) chloride, ≥99.995% trace metals basis
Sigma-Aldrich
Titanium(IV) chloride, ReagentPlus®, 99.9% trace metals basis
Sigma-Aldrich
Phosphoric acid, puriss. p.a., crystallized, ≥99.0% (T)
Sigma-Aldrich
Phosphoric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥85%
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Creatine, anhydrous
Sigma-Aldrich
Phosphoric acid, BioReagent, suitable for insect cell culture, 85%
Sigma-Aldrich
Phosphoric acid, crystalline, ≥99.999% trace metals basis
Sigma-Aldrich
Phosphoric acid, ≥85 wt. % in H2O, ≥99.999% trace metals basis
Sigma-Aldrich
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Phosphoric acid, BioUltra, ≥85% (T)
Sigma-Aldrich
1,1,3,3-Tetraethoxypropane, ≥96%
Sigma-Aldrich
Phosphoric acid-16O4 solution, 70 wt. % in D2O, 99.9 atom % 16O
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Titanium(IV) chloride solution, 1.0 M in methylene chloride
Sigma-Aldrich
Titanium(IV) chloride solution, 1.0 M in toluene
Sigma-Aldrich
Titanium(IV) chloride solution, 0.09 M in 20% HCl
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
2-Thiobarbituric acid, ≥98%
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
4,4′-Diaminodiphenylmethane, ≥97.0% (GC)