  • Search Results
  • Functional similarities between heterogeneously and homogenously expressed MscL constructs.
Functional similarities between heterogeneously and homogenously expressed MscL constructs.
European biophysics journal : EBJ (2015-08-04)
Gamma Chi, Paul R Rohde, Pietro Ridone, Ben Hankamer, Boris Martinac, Michael J Landsberg
ABSTRACT
The mechanosensitive channel of large conductance MscL is a well-characterized mechanically gated non-selective ion channel, which often serves as a prototype mechanosensitive channel for mechanotransduction studies. However, there are some discrepancies between MscL constructs used in these studies, most notably unintended heterogeneous expression from some MscL expression constructs. In this study we investigate the possible cause of this expression pattern, and compare the original non-homogenously expressing constructs with our new homogeneously expressing one to confirm that there is little functional difference between them. In addition, a new MscL construct has been developed with an improved molar extinction coefficient at 280 nm, enabling more accurate protein quantification.
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetonitrile
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 40.0% 2-propanol, 0.05% formic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
SAFC
HEPES
SAFC
HEPES
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 0.1 % (v/v) formic acid, 5 % (v/v) water, suitable for HPLC